GeneAmp™ dNTP Blend (100 mM)
Applied Biosystems™

GeneAmp™ dNTP Blend (100 mM)

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP).
Have Questions?
Catalog NumberQuantity
N80802611 mL
Catalog number N8080261
Price (USD)
913.65
Online exclusive
1,004.00
Save 90.35
Each
Quantity:
1 mL
Request bulk or custom format
Price (USD)
913.65
Online exclusive
1,004.00
Save 90.35
Each

GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial contains a blend of 100 mM solution of dNTP (25 mM each of dATP, dCTP, dGTP and dTTP). Sufficient for 2000 PCR amplifications, 50 μL each. Each kit contains 12 x 1 mL vials.

Key advantages of GeneAmp dNTPs

  • High purity dNTPs are ideal for use in PCR reactions or other applications
  • Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
  • Premixed dNTP blends are available in various concentrations, offering ease of use

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineGeneAmp
Quantity1 mL
Shipping ConditionDry Ice
Concentration100 mM
For Use With (Application)Standard PCR
FormLiquid
Unit SizeEach
Contents & Storage
GeneAmp dNTP Blend, 100 mM (1 mL)

Store in a constant temperature freezer at –20°C.

Frequently asked questions (FAQs)

Why do some dNTP mixes contain dTTP and others TTP?

Thymine is normally found as a deoxyribose. Thus, TTP and dTTP are essentially the same thing. Both types of mixes can be used in PCR reactions