Thermo Scientific™

Lab Vision™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody

Related applications:

Anatomical Pathology

Take advantage of a quality product that delivers accurate results with the Thermo Scientific™ Negative Control for Rabbit IgG Ab-1, Rabbit Polyclonal Antibody.
Back to top
This IgG was purified from serum collected from a rabbit prior to immunization. It does not react with any antigen of human or animal cells. The serum was purified under the same experimental conditions as for the other rabbit polyclonal antibodies of NeoMarkers. In order to serve as a true negative control, this IgG should be used under the identical conditions and at the same concentration as NeoMarkers' other purified rabbit polyclonal IgG antibodies.

Host Species: Rabbit

Isotype: IgG

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO