Thermo Scientific™

Lab Vision™ Negative Control for Mouse IgG1 Ab-1, Mouse Monoclonal Antibody

 Related applications:
Anatomical Pathology
Provides accurate, reproducible results with the Thermo Scientific™ Negative Control for Mouse IgG1 Ab-1, Mouse Monoclonal Antibody.

Error loading your content!

  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart

{{addItemToList.successMessage}} {{addItemToList.listName}}


Back to top

Description

This mouse monoclonal IgG1 was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG1 does not react with any antigen of human or animal cells. This monoclonal IgG1 was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG1 should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG1 antibodies.

Host Species: Mouse

Clone: NCG01

Isotype: IgG1/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO



Documents

Manuals & protocols