Specifications
Certifications/ComplianceAll NanoDrop instruments are approved to CE and UL/CSA.
CompatibilityDYMO LabelWriter 450 printer, Bluetooth keyboard, mouse and barcode reader
ConcentrationdsDNA: Pedestal: 27,500 ng/μL
BSA (IgG): Pedestal: 820 (400) mg/mL
ConnectionsThree USB-A ports and Ethernet enabled
Detection RangedsDNA: Pedestal: 2.0 ng/μL, Cuvette: 0.2 ng/μL,
BSA (IgG): Pedestal: 0.06 (0.03) mg/mL, Cuvette: 0.006 (0.003) mg/mL
Dimensions (L x W x H)20 x 25.4 x 32.3 cm (8 x 10 x 12.7 in.)
For Use With (Application)Nucleic Acid A260, A260/A280, A260/A230 and Labeled Nucleic Acids;, Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Labeled Proteins,, OD600, Kinetics, UV-Vis, and Custom Methods
IncludesND OneC Spec, Wi-Fi
KeypadBuilt-in touchscreen, multipoint capacitive touch
Measurement Time8 seconds
Pathlength0.030 to 1.0 mm (auto-ranging)
Photometric Accuracy3% at 0.97 A, 302 nm, (Absorbance expressed at Abs/mm at 25°C)
Photometric Range10 mm equivalent: Pedestal: 0–550 A; Cuvette: 0–1.5 A
Power ConsumptionOperating: 12 to 18 W; Standby: 5 W
ProcessorAndroid™
RepeatabilityTypical: 0.002 A (1.0 mm path) or 1%CV, whichever is greater
(SD of 10 individual measurements at 0.97 A)
Sample TemperatureTemperature Control (cuvette only) 37 °C
Sample Volume (Metric)Minimum 1μL
System RequirementsWindows™ 8.1 and 10, 64 bit
Wavelength Accuracy±1 nm
Weight (English)7.9 lb.
Weight (Metric)3.6 kg
Detector Type2048-element CMOS Linear Image Sensor
Display7 in. High Definition Color Display, 1280 x 800 pixels
LanguagesEnglish, French, German, Spanish, Polish, Chinese, Japanese, Korean
Light SourceXenon Flash
Spectral Bandwidth≤1.8 nm (FWHM at Hg 254 nm)
Voltage12 VDC
Wavelength Range190 to 850 nm
Unit SizeEach