Achieve superior Fluorescence accuracy and sensitivity for NanoDrop™ Ultra dsDNA BR Assay Kit  and NanoDrop™ Ultra RNA BR Assay Kit quantification across 10–1,000 ng/μL, while minimizing interference from contaminants. Maintain peak performance with the NanoDrop™ Ultra FL-1 Fluorescence Verification Kit for routine fluorescence validation.
NanoDrop Ultra Microvolume UV-Vis Spectrophotometers and Fluorometers
Thermo Scientific™

NanoDrop Ultra Microvolume UV-Vis Spectrophotometers and Fluorometers

The Thermo Scientific™ NanoDrop™ Ultra Spectrophotometer and Fluorometers empower scientists to quantify and assess nucleic acids and proteins with enhanced precision, speed, and reliability.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludesSample Volume (Metric)Spectral Bandwidth
NDULTRACFLGLND UltraC FL, Spec/FluorometerAbs: Min 1μL FL: Min 2μL≤1.8 nm (FWHM at Hg 254 nm)
NDULTRAGLND Ultra Microvolume SpecMin 1μL1.8 nm
NDULTRACGLND UltraC Microvolume SpecMin 1μL≤1.8 nm (FWHM at Hg 254 nm)
NDULTRAFLGLND Ultra FL, Spec/FluorometerAbs: Min 1μL FL: Min 2μL≤1.8 nm (FWHM at Hg 254 nm)
Catalog number NDULTRACFLGL
Price (USD)
17,550.00
Each
Includes:
ND UltraC FL, Spec/Fluorometer
Sample Volume (Metric):
Abs: Min 1μL FL: Min 2μL
Spectral Bandwidth:
≤1.8 nm (FWHM at Hg 254 nm)
Price (USD)
17,550.00
Each

The Thermo Scientific™ NanoDrop™ Ultra Spectrophotometers and Fluorometers represent the next generation of microvolume detection. These cutting-edge instruments deliver exceptional accuracy and sensitivity for nucleic acid and protein analysis with sample volumes as low as 1.0 μL. The NanoDrop Ultra is designed to streamline workflows across a wide range of fields, including genomics, proteomics, molecular biology, biotechnology, pharmaceutical, and life sciences research. It provides unparalleled precision, efficiency, and reliability in one compact device.

New NanoDrop™ Ultra Fluorescence Assays are now available for NanoDrop Ultra FL and UltraC FL instruments. The NanoDrop™ Ultra dsDNA BR Assay Kit (Cat. No. NDU-FL-DSDNA-BR) and NanoDrop™ Ultra RNA BR Assay Kit (Cat. No. NDU-FL-RNA-BR) deliver highly accurate, interference-free quantification across a broad 10-1,000 ng/μL range–offering greater sensitivity and reliability than traditional UV absorbance methods while preserving precious samples for downstream applications. The NanoDrop™ Ultra FL-1 Fluorescence Verification Kit (Cat. No. CHEM-FL1-KIT) helps maintain optimal fluorometer performance with multi-wavelength verification for routine quality assurance. Expand your NanoDrop Ultra workflow with enhanced precision, confidence, and compliance-ready performance.

Avoid costly setbacks in your workflow with an understanding of sample quality like never before using the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers and Fluorometers. Innovative Thermo Scientific™ Acclaro™ Sample Intelligence technology is built into every instrument for improved measurement accuracy and contaminant identification. Quantify and qualify DNA, RNA, and protein samples in seconds with only 1-2μL, and obtain full-spectral data before you decide to use samples in downstream applications. Cuvette compatible models are also available for additional flexibility.

Paired with its compact design and large high-resolution 10.1 in. touch screen interface, the NanoDrop Ultra spectrophotometers are a perfect fit for users looking for flexibility and convenience. The optional Acclaro Pro software (Cat. No. ND-Ultra-AP-GL), compatible with the NanoDrop Ultra instruments, unlocks elevated accuracy for highly concentrated samples with an improved algorithm. This is the next generation NanoDrop single channel instrument with improved hardware and software features over the beloved NanoDrop 2000/2000c and NanoDrop One/OneC Spectrophotometers.

Acclaro Sample Intelligence Technology

Acclaro technology enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants can prevent failure of downstream applications and save days of troubleshooting.

  • Contaminant detection and analysis – offers a comprehensive evaluation of sample purity AND corrects sample absorbance, empowering users to confidently identify, quantify, and mitigate contaminants. Plant and bacteria DNA and RNA differentiations are available for researchers, scientists, and professionals in fields such as agriculture, plant biology, microbiology, and genetic engineering.
  • Sample integrity insight - provides real-time alerts when bubbles are detected in your sample which could otherwise lead to inaccurate measurements.
  • Reliable accuracy – ensures measurement integrity with embedded sensor and digital image analysis.

Tools to aid in 21 CFR Part 11 compliance

Optional Thermo Scientific™ SciVault™ 2 software installs on either the instrument or a companion PC and gives labs the tools they need to comply with US FDA 21 CFR Part 11.

  • Compliance made easy – SciVault 2 (Cat. No. NDU-SCIVAULT2) integrates into the NanoDrop Ultra software user interface and controls user account management, user account access, digital signatures, and audit logs.
  • On-board options – software is available to be run directly on the local instrument through on-board user account management or you can connect your instrument to your domain to use your Windows account management.
  • Centralized solution - install SciVault 2 software on a central computer to control privileges and view audit logs across multiple NanoDrop Ultra instruments in different labs.

Streamlined workflow

  • Fully integrated qPCR recipe calculator - use the built-in library of kits, or add your favorite, to let the software run the math for you. The qPCR recipe calculator suggests how to prepare the reaction based on your sample's concentration.
  • Auto-Measure and Auto-Blank functions – multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.

Advanced connectivity for data storage, printing and sharing

  • Export to the cloud – export data directly to popular cloud storage services like Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
  • Direct LIMS integration – send data directly into your LIMS with the assistance of an integrated RESTful API.
  • Easy data-to-printer options – Print your results directly from the NanoDrop Ultra instrument via Wi-Fi, Ethernet or USB printer connection; print exported data from your PC using the Nanodrop Ultra PC Control software.
  • Data Your Way – customize and preview the information included in the data file prior to export to ensure it meets any downstream needs.

Accuracy beyond expectations with the Acclaro Pro Software (Cat. No. ND-Ultra-AP-GL)

This optional software package includes new versions of the most popular measurement applications which are unlocked from the factory, providing ±5% Error throughout the absorbance dynamic range.

  • Accurate high concentration measurements – absorbance measurements within 5% error up to 550 Abs (10 mm equivalent) – up to 400 mg/mL IgG or 18,000 ng/μL.
  • Quick dilution-free process – take triplicate measurements on the pedestal in 2 minutes.
  • Enhanced calibration – bundled purchases get an additional calibration from the factory to calibrate the instrument for exceptional accuracy within the Acclaro Pro applications.

For research use only; not for diagnostic purposes.

Specifications
Certifications/ComplianceAll NanoDrop instruments are approved to CE and UL/CSA.
CompatibilityDYMO 550 Turbo printer, Bluetooth keyboard, mouse and barcode reader
ConcentrationdsDNA: Pedestal: 27,500 ng/μL BSA (IgG): Pedestal: 820 (400) mg/mL
Connections2x USB-A Ports, 1x USB-C Port, Ethernet, Bluetooth™, Wi-Fi
Detection RangedsDNA: Pedestal: 1.0 ng/μL, Cuvette: 0.2 ng/μL, BSA (IgG): Pedestal: 0.03 (0.015) mg/mL, Cuvette: 0.006 (0.003) mg/mL
Dimensions (L x W x H)12.6 x 7.1 x 11.0 in (32 x 18 x 28 cm)
For Use With (Application)Nucleic Acid A260, A260/A280, A260/A230 and Labeled Nucleic Acids, Oligos;, Protein A280 and A205, Protein Pierce 660, Protein Bradford, Protein BCA, Protein Lowry, Labeled Proteins, dsDNA Fluorescence, RNA Fluorescence, Fluorometer, OD600, Kinetics, UV-Vis, and Custom Methods, Acclaro Pro Applications
IncludesND UltraC FL, Spec/Fluorometer
KeypadBuilt-in touchscreen
Measurement TimeStandard: ≤7 sec Acclaro Pro: ≤30 sec
Pathlength0.030 to 1.0 mm (auto-ranging)
Photometric Accuracy3% at 0.97 A, 302 nm, (Absorbance expressed at Abs/mm at 25°C)
Photometric Range10 mm equivalent: Pedestal: 0-550 A, Cuvette: 0-1.5 A
Power ConsumptionOperating: 15 W; Standby: 7 W
ProcessorCustom Linux
RepeatabilityTypical: 0.002 A (1.0 mm path) or 1%CV, whichever is greater (SD of 10 individual measurements at 0.97 A)
Sample TemperatureTemperature Control (cuvette only) 37°C
Sample Volume (Metric)Abs: Min 1μL FL: Min 2μL
System RequirementsWindows 10 or Windows 11
Warranty2 Years
Wavelength Accuracy±1 nm
Weight (English)Approximately 9.0 Ib.
Weight (Metric)4.1 kg
Detector Type2048-element CMOS Linear Image Sensor
Display10 in. High Definition Color Display
LanguagesChinese, English, French, Japanese, German, Spanish, Polish, Italian, Korean
Light SourceXenon Flash, Blue LED, Red LED
Product LineNanoDrop
Spectral Bandwidth≤1.8 nm (FWHM at Hg 254 nm)
Voltage12 VDC
Wavelength Range190 to 850 nm
Unit SizeEach

Frequently asked questions (FAQs)

How do I obtain service for my NanoDrop Ultra?

For service in the United States and Canada, please call 877-724-7690 option 3 or email nanodrop@thermofisher.com.
For international service, please contact your local NanoDrop distributor.

Am I able to connect my LIMS platform to NanoDrop Ultra?

All NanoDrop Ultra instruments come with an integrated RESTful API. Please refer to the NanoDrop Ultra API Reference Manual, available on our website, for more information on how to connect to and use the API. Visit www.thermofisher.com/nanodrop.

Does NanoDrop Ultra come with 21 CFR Part 11 compliance software?

Optional SciVault 2 Software installs on either the instrument or a companion PC and gives labs the tools they need to comply with US FDA 21 CFR Part 11. This software allows NanoDrop Ultra users to manage user accounts, control user account access, apply digital signatures, and maintain audit logging.

What is Acclaro Pro?

Acclaro Pro is an optional enhanced software package for NanoDrop Ultra. It incorporates advanced algorithms and expanded measurement applications designed for higher accuracy (e.g., ±5% error across a wider dynamic range) and better performance with highly concentrated or regulated samples. This is especially useful in biopharma, QC, and manufacturing contexts where tighter quantitation and broader applications are needed.

What is the difference between the NanoDrop Ultra and NanoDrop Ultra FL?

The NanoDrop UltraC includes both the standard microvolume pedestal and an additional cuvette port for measuring dilute or larger-volume samples, whereas the NanoDrop Ultra is pedestal-only.