Bolt™ Bis-Tris Plus Resupply Pack, 4 to 12%, WedgeWell™ format
Invitrogen™

Bolt™ Bis-Tris Plus Resupply Pack, 4 to 12%, WedgeWell™ format

The Bolt Bis-Tris Plus Resupply Pack optimizes your protein separation workflow. This pack includes precast gels, running buffer, and protein ladders, enough for 100 gel runs.
Have Questions?
變更視圖buttonViewtableView
產品號碼Wells
NW0412A-RS1010-well
NW0412B-RS1012-well
NW0412C-RS1015-well
產品號碼 NW0412A-RS10
價格 (TWD)
77,770.00
線上優惠
Ends: 30-Jun-2026
111,100.00
您節省 33,330.00 (30%)
Each
Wells:
10-well
價格 (TWD)
77,770.00
線上優惠
Ends: 30-Jun-2026
111,100.00
您節省 33,330.00 (30%)
Each
The Bolt Bis-Tris Plus Resupply Pack optimizes your protein separation workflow. This pack includes precast gels, running buffer, and protein ladders, enough for 100 gel runs. Bolt Bis-Tris Plus gels offer the fastest Bis-Tris gel run time, superior broad-range separation, and maintain protein integrity due to their neutral pH environment. Designed for optimal separation of diverse protein sizes under denaturing conditions, they are ideal for western blot transfer and analysis. With a long 16 month shelf life and the innovative WedgeWell format for easier sample loading, this pack is a reliable and convenient solution for your laboratory needs.

The Bolt Bis-Tris Plus Resupply Pack contains:

  • Bolt Bis-Tris Plus gels, WedgeWell format (10 boxes, 100 gels)
  • Bolt MES Running Buffer for 100 gel runs, 20X (4 bottles, Cat. No. B0002)
  • PageRuler Plus Pre-stained Protein Ladder for 100 gel runs, 10 to 250 kDa (Cat. No. 26619)

About Bolt Bis-Tris Plus Gels

Bolt Bis-Tris Plus gels are high-performance precast polyacrylamide gels designed to provide optimal broad range separation of proteins under denaturing conditions. These gels operate at a neutral pH, minimizing protein modifications and ensuring protein integrity, making them ideal for western blot transfer and analysis. The unique WedgeWell design allows for up to 2X more sample volume loading than conventional precast gels, facilitating detection of proteins in extremely diluted samples or visualization of low-abundance proteins. Bolt Bis-Tris Plus Gels, combined with Bolt LDS Sample Buffer and the Invitrogen NuPAGE Antioxidant running buffer additive, deliver sharp, clear protein bands, preserving protein integrity throughout electrophoresis. Stable at room temperature, these gels are a practical and efficient choice for your lab.

Separation

For running Invitrogen protein gels, you have the flexibility to use either the Mini Gel Tank (A25977) or the XCell SureLock Mini-Cell (EI0001). Both systems are designed to provide optimal performance. The Mini Gel Tank's unique side-by-side design offers efficient and convenient electrophoresis for both precast and SureCast handcast mini gels. Alternatively, the XCell SureLock Mini-Cell allows quick, easy, and leak-free running of mini gels, without the need for any clamps. This compatibility gives you the freedom to choose the best tool for your specific lab and experiment requirements. Additionally, the Mini Gel Tank is conveniently available through Invitrogen Precast Gel Welcome Packs along with both sample buffer and reducing agent, providing an all-in-one solution for your electrophoresis needs.

Transfer

For transferring your proteins to a membrane, we recommend using for Bolt Transfer Buffer (BT0006) for traditional wet transfer using the Mini Blot Module (B1000). Alternatively, rapid semi-dry transfer can be done using the Invitrogen Power Blotter or rapid dry transfer using the iBlot 2 Gel Transfer Device (IB21001).

For Research Use Only. Not for use in diagnostic procedures.
規格
For Use With (Equipment)XCell SureLock Mini-Cell
Quantity1 Kit
Gel Percentage4 to 12%
Gel SizeMini
Gel TypeBis-Tris Plus
Separation Range3.5 to 260 kDa
Separation TypeDenaturing
Separation ofProtein
Wells10-well
Unit SizeEach
內容物與存放
• Bolt Bis-Tris Plus gels, 10 x 10 gels
• Bolt MES Running Buffer (20X), 4 bottles
• PageRuler Plus Pre-stained Protein Ladder (10 to 250 kDa)