Get the sharpest, brightest images. Try new Invitrogen™ ProLong™ Glass antifade mountant with glass-matching refractive index of 1.52. Learn more

ONCYTE™ MultiWells, one well, 13 mm diameter, with slide & matching gasket

Catalog number:  O24751

 Related applications: Cellular Imaging

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The versatile ONCYTE® MultiWells consist of a two-piece set that includes a slide printed with nitrocellulose circles and a matching removable gasket to enclose and isolate each sample. The nitrocellulose coating on the slide is specially formulated for fluorescent imaging. The ultra-thin microporous coating ensures uniform binding of tissue prints, cells or macromolecules and becomes transparent in a variety of mounting media. The matching press-to-seal silicone gaskets adhere easily to the surface of the slide to isolate specimens and reagents and prevent cross contamination.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Dimensions: 13 mm diameter
Product Line: ONCYTE®
Product Size: 1 set
Shipping Condition: Room Temperature
Type: Gaskets, Slides

Contents & storage

Store at room temperature.

Documents

Manuals & protocols