Phalloidin Labeling Probes
Invitrogen™

Phalloidin Labeling Probes

Achieve precise and reliable F-actin staining with fluorescent and biotinylated phalloidins. Phalloidin conjugates are widely used in imaging applications to selectively label F-actin in a variety of sample types including fixed and permeabilized cells, tissue sections, and cell-free experiments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorExcitation Wavelength RangeDye Type
O7466Green496/520 nmOregon Green™ 488
A22281Blue346/442 nmAlexa Fluor™ 350
A30104Violet405/450 nmAlexa Fluor™ Plus 405
A12379Green495/518 nmAlexa Fluor™ 488
F432Green496/516 nmFITC (Fluorescein)
A22282Yellow531/554 nmAlexa Fluor™ 532
R415Red-orange540/565 nmTRITC
A22283Orange556/570 nmAlexa Fluor™ 546
A34055Orange555/565 nmAlexa Fluor™ 555
A30106Orange555/565 nmAlexa Fluor Plus 555
B3475Red558/569 nmBODIPY™
A12380Orange-red578/600 nmAlexa Fluor™ 568
A12381Red581/609 nmAlexa Fluor™ 594
T7471Red591/608 nmTexas Red™
A22284Far-red632/647 nmAlexa Fluor™ 633
A34054Far-red633/647 nmAlexa Fluor™ 635
A22287Far-red650/668 nmAlexa Fluor™ 647
A30107Far-red650/668 nmAlexa Fluor Plus 647
A22285Near-infrared663/690 nmAlexa Fluor™ 660
A22286Near-infrared679/702 nmAlexa Fluor™ 680
A30105Near-infrared758/784 nmAlexa Fluor™ Plus 750
B7474NoneNoneBiotin-XX
P3457NoneNonePhalloidin (unlabeled)
Catalog number O7466
Price (USD)
609.65
Online exclusive
656.00
Save 46.35
Each
Color:
Green
Excitation Wavelength Range:
496/520 nm
Dye Type:
Oregon Green™ 488
Price (USD)
609.65
Online exclusive
656.00
Save 46.35
Each
Fluorescent and biotinylated phalloidins are water soluble and bind to filamentous actin (F-actin) with nanomolar affinity, making them convenient probes for labeling, identifying, and quantifying F-actin in cryopreserved tissue sections, fixed and permeabilized cells, and cell-free experiments. Phalloidin conjugates bind similarly to actin from various species, including plants and animals, enabling staining of the cytoskeleton in a wide range of samples.

A variety of phalloidin conjugates for filamentous (F-actin) staining are available, including fluorescent Alexa Fluor and Alexa Fluor Plus phalloidins, along with phalloidins conjugated to classic fluorescent dyes such as BODIPY, fluorescein, and rhodamine. Phalloidin staining is spectrally compatible with other fluorescent stains used in cellular analyses such as GFP/RFP, Qdot nanocrystals, and other Alexa Fluor conjugates and antibodies. Biotin‐XX Phalloidin can be used to visualize actin filaments via fluorescent streptavidin tags or standard enzyme-mediated avidin/streptavidin techniques such as in electron microscopy. Unlabeled phalloidin is available for use as a control in blocking F‐actin staining or in promoting polymerization.

Phalloidin conjugates bind to both large and small actin filaments with similar affinity in a 1:1 stoichiometry between phallotoxin and actin subunits. They do not bind G-actin monomers.

Alexa Fluor and Alexa Fluor Plus phalloidin conjugates for F-actin staining

Fluorescent Alexa Fluor dye conjugates of phalloidin are popular F-actin stains, offering color choices across the full spectral range. These phalloidin conjugates provide researchers with fluorescent probes that are superior in brightness and photostability compared to other spectrally similar conjugates.

Alexa Fluor Plus Phalloidin conjugates retain the same specificity for actin but offer 3-5 times greater sensitivity and brightness compared to the corresponding Alexa Fluor Phalloidin conjugate. This increased brightness is beneficial for challenging F-actin imaging, such as the super‐resolution microscopy methods SIM and STORM, and for reliable staining of actin stress fibers.

Features of phalloidin probes

  • High specificity—binds selectively to F-actin, which allows for precise labeling of actin filaments in fixed cells and cryopreserved tissues
  • Strong affinity—nanomolar binding affinity for F-actin, which ensures stable and reliable actin staining
  • Extensive fluorescent conjugate options—over twenty conjugated varieties of phalloidin
  • Compatibility with fixed samples—typically used with fixed cells and tissues, making them suitable for actin staining in detailed structural studies, immunofluorescence staining, and IHC applications
  • Multiplexing capability—the wide availability of phalloidin conjugates enables their use in combination with other fluorescent probes and antibodies for multiplex imaging. Biotinylated phalloidin can be made use of in downstream streptavidin steps.
  • Quantitative analysis—can be used for quantitative analysis of F-actin distribution and density within cells, aiding in the study of cytoskeletal dynamics. The unlabeled phalloidin can be titrated as a control.
  • Ease of use—staining is straightforward and quick
  • Excellent stability—exhibit good photostability, which is essential for prolonged imaging sessions and time-lapse studies
  • Wide applicability—used for a range of applications, including studying cell morphology, motility, and the effects of drugs on the actin cytoskeleton
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorGreen
Dye TypeOregon Green™ 488
Excitation Wavelength Range496/520 nm
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Confocal Microscope, Compatible with FITC/GFP filter set
Product LineOregon Green
Quantity300 Units
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Product TypePhalloidin
Sub Cellular LocalizationActin, Cytoskeleton
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Frequently asked questions (FAQs)

Do you offer alternative package sizes for phalloidin or ReadyProbes reagents?

Alternative package sizes may be available through a custom request process. Contact Thermo Fisher Scientific customer support or your sales representative to discuss available options for the specific reagent and quantity needed.

Can I use conjugated phalloidin label probes on organoids?

Yes. Conjugated phalloidin labeling probes can be used with organoids when F-actin is accessible after fixation and permeabilization. Because organoids are thicker than monolayer cell cultures, protocol optimization may be needed, including permeabilization conditions, probe incubation time, and probe concentration.

Are conjugated phalloidin reagents antibodies?

No, Phalloidin is not an antibody. It is a bicyclic peptide toxin originally isolated from the Amanita phalloides mushroom and is used to bind and label F-actin in fixed and permeabilized samples.

Can I use methanol instead of DMSO to reconstitute the conjugated phalloidins from the lyophilized solids?

Yes, methanol can be used to reconstitute fluorescent phalloidin, but it creates a less concentrated stock solution than DMSO. Dissolve the vial contents in 1.5 mL of methanol to yield a 40X stock solution at a concentration of 200 assays/mL, which is equivalent to approximately 6.6 µM. One unit/assay of fluorescent phalloidins is equivalent to 5 µL of the methanolic stock solution. Methanol may be used as a stock-solution solvent, but methanol-containing fixatives are not recommended because they can disrupt F-actin.

How do I reconstitute fluorescent conjugated phalloidin probes to make a stock solution?

For fluorescent phalloidin label probes, prepare a DMSO stock solution by dissolving the vial contents in 150 µL anhydrous DMSO to make a 400X stock, with a concentration of approximately 66 µM or 2,000 assays per mL. Store between -25°C and -5°C, protected from light. One unit/assay of fluorescent phalloidins is equivalent to 0.5 µL of the DMSO stock solution.

Citations & References (39)

Citations & References
Abstract
Temporal and spatial distribution of activated Pak1 in fibroblasts.
Authors:Sells MA, Pfaff A, Chernoff J
Journal:J Cell Biol
PubMed ID:11134074
'p21-activated kinases (Paks) are effectors of the small GTPases Cdc42 and Rac, and are thought to mediate some of the cytoskeletal and transcriptional activities of these proteins. To localize activated Pak1 in cells, we developed an antibody directed against a phosphopeptide that is contained within the activation loop of Pak1. ... More
Apical, lateral, and basal polarization cues contribute to the development of the follicular epithelium during Drosophila oogenesis.
Authors:Tanentzapf G, Smith C, McGlade J, Tepass U
Journal:J Cell Biol
PubMed ID:11076972
'Analysis of the mechanisms that control epithelial polarization has revealed that cues for polarization are mediated by transmembrane proteins that operate at the apical, lateral, or basal surface of epithelial cells. Whereas for any given epithelial cell type only one or two polarization systems have been identified to date, we ... More
Modulation of acto-myosin contractility in skeletal muscle myoblasts uncouples growth arrest from differentiation.
Authors:Dhawan J, Helfman DM
Journal:J Cell Sci
PubMed ID:15252113
'Cell-substratum interactions trigger key signaling pathways that modulate growth control and tissue-specific gene expression. We have previously shown that abolishing adhesive interactions by suspension culture results in G(0) arrest of myoblasts. We report that blocking intracellular transmission of adhesion-dependent signals in adherent cells mimics the absence of adhesive contacts. We ... More
Lipid rafts in the maintenance of synapses, dendritic spines, and surface AMPA receptor stability.
Authors:Hering H, Lin CC, Sheng M
Journal:J Neurosci
PubMed ID:12716933
'Cholesterol/sphingolipid microdomains (lipid rafts) in the membrane are involved in protein trafficking, formation of signaling complexes, and regulation of actin cytoskeleton. Here, we show that lipid rafts exist abundantly in dendrites of cultured hippocampal neurons, in which they are associated with several postsynaptic proteins including surface AMPA receptors. Depletion of ... More
ADP-ribosylation factor 6 regulates a novel plasma membrane recycling pathway.
Authors:Radhakrishna H, Donaldson JG
Journal:J Cell Biol
PubMed ID:9314528
'ADP-ribosylation factor (ARF) 6 localizes to the plasma membrane (PM) in its GTP state and to a tubulovesicular compartment in its GDP state in HeLa cells that express wild-type or mutant forms of this GTPase. Aluminum fluoride (AlF) treatment of ARF6-transfected cells redistributes ARF6 to the PM and stimulates the ... More