MAS™ Omni·CORE™ Max Controls
Thermo Scientific™
MAS™ Omni·CORE™ Max Controls
Consolidates general chemistry and serum protein QC processing into a single product offering three distinct levels in a load and go barcoded tube format.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
OCRM-1011
OCRM-2022
OCRM-3033
Catalog number OCRM-101
Price (USD)
-
Level:
1
Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific MAS Omni·CORE Max Controls. Value assignment is provided for key integrated instrument systems. Load-and-Go barcoded1 tubes for improved efficiency and daily time savings 1Barcodes available for selected instruments.
  • Three distinct levels
  • Liquid, ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
Available in select countries only
For In Vitro Diagnostic Use.  
Specifications
DisposableReusable
Level1
SterilityNon-sterile
TypeChemistry Control
FormLiquid
Sample TypeHuman Serum
Unit SizeEach