MAS™ Omni·IMMUNE™ PRO Immunoassay Controls
Actual product may vary
Thermo Scientific™
MAS™ Omni·IMMUNE™ PRO Immunoassay Controls
Consolidate your immunoassay and tumor marker controls into a single QC product with MAS Omni·IMMUNE PRO Immunoassay Controls.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLevel
OPRO-1011
OPRO-2022
OPRO-3033
Catalog number OPRO-101
Price (USD)
499.00
Each
Level:
1
Price (USD)
499.00
Each
Consolidate your immunoassay and tumor marker controls into a single QC product with Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO Immunoassay Controls. This QC product contains the same analyte panel as the Omni·IMMUNE control but also has the following additional analytes: Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO) and Sex Hormone Binding Globulin (SHBG).
  • Liquid, ready-to-use
  • Human-based serum matrix
  • Value assignment provided for most major analyzers
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For In Vitro Diagnostic Use.  
For In Vitro Diagnostic Use.  
Specifications
DescriptionOmni IMMUNE PRO Control, Assayed
Level1
Packaging6 Pk.
Quantity5 mL
Shelf Life36 months at -25° to -15°C
TypeControl
Detectable Analytes17-OH-Progesterone, 25-OH-Vitamin D, Acetaminophen, Adrenocorticotropic Hormone (ACTH), Aldosterone, alpha-Fetoprotein (AFP), Amikacin, Anti-Tg, Anti-TPO, Benzodiazepines, beta-2-microglobulin (B2M), β-hCG, CA-125, CA 15-3, CA 19-9, Carbamazepine, Carcinoembryonic Antigen (CEA), Cortisol, C-peptide, DHEA Sulfate, Digoxin, Disopyramide, Estradiol, Estriol (Free), Ethosuximide, Ferritin, Folate (Vitamin B9), Fructosamine, Follicle Stimulating Hormone (FSH), Gastrin, Gentamicin, Growth Hormone, hCG, Homocysteine, IgE, Insulin-like Growth Factor 1 (IGF-1), Insulin, Luteinizing Hormone (LH), Lidocaine, Lithium, N-acetylprocainamide (NAPA), Prostatic Acid Phosphatase (PAP), Phenobarbital, Phenytoin, Phenytoin (Free), Primidone, Procainamide, Procalcitonin (PCT), Progesterone, Prolactin, PSA (Free), PSA (Total), Parathyroid Hormone (PTH), Quinidine, Salicylate (SALY), Sex Hormone-binding Globulin (SHBG), T3 (Free), Triiodothyroxine (T3 Total), T4 (Free), Thyroxine (T4 Total), Thyroxine-binding Globulin (TBG), Testosterone, Theophylline, Thyroglobulin, Tobramycin, Tricyclic Antidepressants, Thyroid Stimulating Hormone (TSH), T-uptake, Valproic Acid, Valproic Acid (Free), Vancomycin, Vitamin B12
Sample TypeSerum
Unit SizeEach
Contents & Storage
-25°C to -15°C