MAS™ Omni·IMMUNE™ PRO Max Controls
MAS™ Omni·IMMUNE™ PRO Max Controls
MAS™ Omni·IMMUNE™ PRO Max Controls
Provides Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO) and Sex Hormone Binding Globulin (SHBG) in addition to the same analyte panel provided in the MAS Omni·IMMUNE product in a barcoded load and go tube format.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoNivel
OPROM-2022
OPROM-1011
OPROM-3033
Número de catálogo OPROM-202
Precio (MXN)
-
Solicite un presupuesto
Nivel:
2
Consolidate your immunoassay and tumor marker controls into a single QC product with Thermo Scientific MAS Omni·IMMUNE PRO Max Immunoassay Controls. This QC product contains the same analyte panel as the Omni·IMMUNE control but also has the following additional analytes: Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO) and Sex Hormone Binding Globulin (SHBG). Value assignment is provided for key integrated instrument systems. Load-and-Go barcoded1 tubes for improved efficiency and daily time savings 1Barcodes available for selected instruments.
  • Three distinct levels
  • Liquid, ready-to-use
  • Human-serum based matrix
  • Assayed values available for most analyzers
Available in select countries only
Para uso diagnóstico in vitro
Especificaciones
DesechablesReusable
Nivel2
EsterilidadNon-sterile
TipoClinical Chemistry Control
FormularioLiquid Ready-to-Use
Tipo de muestraHuman Serum
Unit SizeEach