Tox Explorer™ Collection
Tox Explorer™ Collection
Tox Explorer™ Collection
Tox Explorer™ Collection
Tox Explorer™ Collection
Thermo Scientific™
Tox Explorer™ Collection
Step up productivity, reduce costs, and ensure high quality results with proven toxicology LC-MS/MS methods, regardless of analyte, matrix complexity, or expertise.
Have Questions?
Catalog NumberType
OPTON-31039Software
Catalog number OPTON-31039
Price (USD)
-
Type:
Software
Be confident with a proven toxicology method and robust LC-MS/MS platform for concise, comprehensive drug analysis. Optimized for quick start-up and ease of use, the Thermo Scientific™ Tox Explorer™ Collection includes easy-to-follow sample preparation guidelines, LC-MS analytical method, hardware and software capabilities, HPLC columns, comprehensive mass spectral library, application support, and training for confident, high-quality results, regardless of analyte, matrix, or user expertise level.
Step up productivity, reduce costs, and achieve high quality data using easily implementable and reliable LC-MS/MS toxicology methods for forensic toxicology, workplace testing, sports antidoping, or therapeutic drug research laboratories. Improve efficiency and reduce cost throughout the preparation, separation, detection and analysis of drug compounds. These methods can be applied to multiple biological matrices, including blood, plasma, and urine with unprecedented speed, ease, and confidence.
Easily Implemented Analytical Methods
  • Proven LC-MS/MS method, confirmed with accuracy and precision data, to analyze large toxicology panels in a single run
  • Step-by-step guide for quick start-up
  • LC-MS hardware parameters
  • Comprehensive training and application support
Definitive Drug Analysis
  • High resolution and robust mass accuracy from Thermo Scientific Orbitrap mass analyzer MS systems
  • Thermo Scientific™ Vanquish™ Flex UHPLC can efficiently separate large panels with varying drugs of abuse containing different polarities and hydrophobicities
  • Reproducible retention time offered via Thermo Scientific™ Accucore™ columns
Enhance identification
  • Thermo Scientific™ TraceFinder™ software offers streamlined data processing along with customized reporting
  • Comprehensive spectral library of 1500+ compounds
  • Ability to conduct retrospective analysis without additional experiments
For General Lab Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeThermo Scientific™ Accucore™ Phenyl-Hexyl HPLC Column
For Use With (Equipment)LC-MS System
IncludesSample preparation guidelines, verified method, installation guide, LC-MS hardware parameters, mass spectral library with real-time updates, comprehensive training, application support
Liquid Chromatography SystemThermo Scientific™ Vanquish™ Flex Binary UHPLC System
Mass Spectrometer InterfaceThermo Scientific™ Q Exactive™ Plus Hybrid Quadrupole-Orbitrap Mass Spectrometer, HRAM Data-Dependent Analysis (DD-MS2)
MethodsAcquisition and processing methods, TraceFinder Compound Database, mzVault library
Sample PreparationProcedure to prepare urine, blood, and plasma/serum samples
SoftwareThermo Scientific TraceFinder Software
TypeSoftware
Unit SizeEach