OptiSpray™ PepMap Neo UHPLC Cartridges
OptiSpray™ PepMap Neo UHPLC Cartridges
OptiSpray™ PepMap Neo UHPLC Cartridges
Thermo Scientific™

OptiSpray™ PepMap Neo UHPLC Cartridges

Perform high-resolution reversed-phase separations on a range of biological samples with Thermo Scientific™ OptiSpray PepMap™ Neo UHPLC cartridges.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDiámetro (métrico)Longitud (métrico)
OS75150PN75 μm15 cm
OS75500PN75 μm50 cm
OS150150PN150 μm15 cm
Número de catálogo OS75150PN
Precio (USD)
-
Diámetro (métrico):
75 μm
Longitud (métrico):
15 cm
Separate complex bottom-up proteomics, metabolomics, and lipidomics samples in a wide range of matrices from cell digests to plasma using the Thermo Scientific™ OptiSpray™ PepMap™ Neo UHPLC cartridge columns. In addition to optimized low-flow chromatographic performance, OptiSpray cartridge columns eliminate challenging capillary connections and manual optimization of source positioning.

Along with the Thermo Scientific™ OptiSpray™ Ion Source, OptiSpray PepMap Neo Cartridges provide:

Performance for deep coverage with high precision and accuracy

Optimized fluidics, integrated temperature and sheath gas control, and automated spray positioning yield reproducible, high-quality results when used with or without the FAIMS interface.

Simplicity for users of all experience levels

Plug-and-play cartridge installation and automated spray optimization virtually eliminate manual setup of nano and capillary columns. On-board memory stores cartridge properties, usage data, and previous source settings to quickly achieve optimal ESI performance, every time.

Reliability across columns, users, and LC-MS systems

Enhanced robustness with an inactive spray position between injections for a cleaner MS system while exchangeable emitters extend column lifetime. Automated spray positioning, cartridge memory, and integrated nebulization gas boost run-to-run and intercolumn result reproducibility.

Especificaciones
Formato de columnaOptiSpray
Tipo de columnaReversed Phase
Con desactivación terminalYes
Caudal100 to 1500 nL/min.
Para utilizar conOptiSpray Ion Source
Para utilizar con (aplicación)LC-MS
Presión máxima22,000 psi (1500 bar)
Material de embalajeSpherical, Fully Porous, Ultrapure Silica
Fase estacionariaC18
Área de la superficie450 m2/g
Temperatura60°C
Tipo USPL1
Carga de carbono15%
Diámetro (métrico)75 μm
Longitud (métrico)15 cm
Tamaño de partículas2 μm
Tamaño de poro100 Å
Línea de productosPepMap Neo
pH2 to 8
Unit SizeEach