OptiSpray™ Emitters
OptiSpray™ Emitters
OptiSpray™ Emitters
OptiSpray™ Emitters
Thermo Scientific™
OptiSpray™ Emitters
Thermo Scientific™ OptiSpray™ Emitters are exchangeable electrospray ionization emitters that seamlessly integrate with the Thermo Scientific™ OptiSpray™ Ion Source and Cartridges.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
OSE1535TOptiSpray Tapered Emitter
OSE0835POptiSpray Pulled Emitter
Catalog number OSE1535T
Price (USD)
2,144.00
Pack of 5
Product Type:
OptiSpray Tapered Emitter
Price (USD)
2,144.00
Pack of 5
Thermo Scientific™ OptiSpray™ Emitters are exchangeable electrospray ionization nano and capillary emitters that seamlessly integrate with the Thermo Scientific™ OptiSpray™ Ion Source and Cartridges. OptiSpray emitters provide high sensitivity, precision, and robustness without the challenges presented when handling traditional ESI emitters. In addition, OptiSpray emitters can be readily exchanged, significantly increasing column lifetime. The tapered emitter comes standard with OptiSpray Cartridges while the pulled emitter.
Specifications
For Use WithOptiSpray Ion Source and Cartridges
For Use With (Application)Increasing column lifetime
Product TypeOptiSpray Tapered Emitter
Diameter (Metric)15 μm
Length (Metric)3.5 cm
Unit SizePack of 5