Thermo Scientific™
Oxoid™ VDRL Test Kit
Catalog number: OXDR0525M
Thermo Scientific™
Oxoid™ VDRL Test Kit
Catalog number: OXDR0525M
Thermo Scientific™ Oxoid VDRL Test Kit is a carbon antigen agglutination test used in Syphilis screening for the detection and semi-quantitative assessment of reagin antibodies in serum or plasma.
Catalog Number
OXDR0525M
Unit Size
Each
Quantity
100 Tests/Kit
Price (USD)
Full specifications
Quantity100 Tests/Kit
DescriptionVDRL Test Kit
Detectable Analytesreagin antibodies in serum or plasma
TypeVDRL Test Kit
Unit SizeEach
Catalog Number
Unit Size
Each
Quantity
500 Tests/Kit
Price (USD)
Full specifications
Quantity500 Tests/Kit
DescriptionVDRL Test Kit
Detectable Analytesreagin antibodies in serum or plasma
TypeVDRL Test Kit
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeQuantityPrice (USD)
OXDR0525MFull specifications
Each100 Tests/KitRequest A Quote
Quantity100 Tests/Kit
DescriptionVDRL Test Kit
Detectable Analytesreagin antibodies in serum or plasma
TypeVDRL Test Kit
Unit SizeEach
OXDR0526MFull specifications
Each500 Tests/KitRequest A Quote
Quantity500 Tests/Kit
DescriptionVDRL Test Kit
Detectable Analytesreagin antibodies in serum or plasma
TypeVDRL Test Kit
Unit SizeEach
Showing 2 of 2
Thermo Scientific™ Oxoid VDRL Test Kit is a carbon antigen agglutination test used in Syphilis screening for the detection and semi-quantitative assessment of reagin antibodies in serum or plasma.
Contents & Storage
2°C to 8°C