Search Thermo Fisher Scientific
Thermo Scientific™ Oxoid VDRL Test Kit is a carbon antigen agglutination test used in Syphilis screening for the detection and semi-quantitative assessment of reagin antibodies in serum or plasma.
Description | VDRL Test Kit |
Quantity | 500 Tests/Kit |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
OXDR0526M | Each | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Thermo Scientific™ Oxoid VDRL Test Kit is a carbon antigen agglutination test used in Syphilis screening for the detection and semi-quantitative assessment of reagin antibodies in serum or plasma.