Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
Fisher Chemical™

Phenol Detached Crystals, Certified AR, for Analysis, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™

C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113 | CAS: 108-95-2 | C6H6O | 94.11 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 g
1 kg
產品號碼 P-2360-60
亦稱為 P/2360/60
價格 (TWD)
-
Quantity:
1 kg
Request bulk or custom format
規格
CAS108-95-2
ChEBICHEBI:15882
Chemical Name or MaterialPhenol Detached Crystals
ColorColorless to White
Formula Weight94.11g/mol
檢視更多
C6H6O, CAS Number-108-95-2, oxybenzene, hydroxybenzene, carbolic acid, phenyl hydroxide, monophenol, phenyl hydrate, phenic acid, phenylic acid, phenylic alcohol, benzenol, 1kg, 182 deg.C, CHEBI:15882, Colorless to White, 94.11g/mol, ISWSIDIOOBJBQZ-UHFFFAOYSA-N, 39 deg.C, 2143, 94.113