Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™
Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™
Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™
Fisher Chemical™

Phenolphthalein Solid, Pure, Indicator Grade, Fisher Chemical™

Have Questions?
Catalog NumberQuantity
P-2400-48
also known as P/2400/48
100 g
Catalog number P-2400-48
also known as P/2400/48
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Specifications
CAS77-09-8
ChEBICHEBI:34914
Formula Weight318.32g/mol
IUPAC Name3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
View more
Phenolphthalein Solid, Pure, Indicator Grade