Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™
Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™
Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™

Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 1kg, CAS: 16788-57-1, White, Molecular Formula: H7K2O7P, Molecular Weight: 228.219, Packaging: Plastic powder jar, pH: 8.5 to 9.6
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
Catalog number P-5250-60
also known as P/5250/60
Price (USD)
-
Quantity:
1 kg
Request bulk or custom format
Specifications
CAS16788-57-1
ColorWhite
Formula Weight228.23g/mol
IUPAC Namedipotassium;hydrogen phosphate;trihydrate
InChI KeyXQGPKZUNMMFTAL-UHFFFAOYSA-L
View more
Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 1kg, CAS: 16788-57-1, White, Molecular Formula: H7K2O7P, Molecular Weight: 228.219, Packaging: Plastic powder jar, pH: 8.5 to 9.6