Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Fisher Chemical™

Potassium Hydroxide, Certified AR for Analysis, Pellets, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

HKO, CAS Number-1310-58-3, potash lye, caustic potash, hydroxyde de potassium, caswell no. 693, potassium hydrate, potassium hydroxide, potasse caustique, potassa, potassium hydroxide solution, potassium hydroxide k oh, 500g, 1320 deg.C, CHEBI:32035, White, 56.11g/mol, KWYUFKZDYYNOTN-UHFFFAOYSA-M
Have Questions?
Catalog NumberQuantity
P-5640-53
also known as P/5640/53
500 g
Catalog number P-5640-53
also known as P/5640/53
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point1320°C
CAS1310-58-3
ChEBICHEBI:32035
ColorWhite
Formula Weight56.11g/mol
View more
HKO, CAS Number-1310-58-3, potash lye, caustic potash, hydroxyde de potassium, caswell no. 693, potassium hydrate, potassium hydroxide, potasse caustique, potassa, potassium hydroxide solution, potassium hydroxide k oh, 500g, 1320 deg.C, CHEBI:32035, White, 56.11g/mol, KWYUFKZDYYNOTN-UHFFFAOYSA-M