Pyridine, Certified AR for Analysis, Fisher Chemical™
Pyridine, Certified AR for Analysis, Fisher Chemical™
Pyridine, Certified AR for Analysis, Fisher Chemical™
Fisher Chemical™

Pyridine, Certified AR for Analysis, Fisher Chemical™

C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
Amber Glass Bottle
Catalog number P-7960-08
also known as P/7960/08
Price (TWD)
-
Quantity:
500 mL
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Boiling Point115°C
CAS110-86-1
ChEBICHEBI:16227
ColorColorless
Formula Weight79.1g/mol
View more
C5H5N, CAS Number-110-86-1, azabenzene, azine, caswell no. 717, piridina, piridina italian, pirydyna, pirydyna polish, pyridin, pyridin german, rcra waste number u196, 2.5L, 115 deg.C, CHEBI:16227, Colorless, 79.1g/mol, JUJWROOIHBZHMG-UHFFFAOYSA-N, pyridine, -42 deg.C, 11732, 79.102