RapidINTEL™ Positive Control Cartridge
RapidINTEL™ Positive Control Cartridge
Applied Biosystems™
RapidINTEL™ Positive Control Cartridge
The Applied Biosystems RapidINTEL Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrumentRead more
Have Questions?
Catalog NumberQuantity
P0443961 Cartridge
Catalog number P044396
Price (KRW)
438,000
Each
Add to cart
Quantity:
1 Cartridge
Price (KRW)
438,000
Each
Add to cart
The Applied Biosystems RapidINTEL Positive Control Cartridge is for use with the Applied Biosystems RapidHIT ID System to verify instrument and/or reagent performance. Each cartridge is individually wrapped and single-use/disposable.
Required for use:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
• Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID System
Quantity1 Cartridge
FormatCartridge
Unit SizeEach