Search
Search

| Catalog Number | Type |
|---|---|
| P10-CL | Side Clamps |
| P10-20G | Blank Glass |
| P10-20R | Notched Glass |
| P10DS-001 | Upper Buffer Chamber |
| P10DS-006 | Blocking Plate |
| P10DS-008 | Lower Buffer Chamber |
| P10DS-012 | Lid |
| P2-SC | Spacer, Thin |
| P2-SD | Spacer, Thick |
Keep your dual gel system running with Thermo Scientific™ Owl™ P10DS Dual Gel System Replacement Parts. Parts are for use with the Owl™ P10DS Dual Gel System, an optimal device for a large number of samples that need extended separation and mobility shift assays.