Ethyl Alcohol with Deionized Water, 140 proof
Greenfield Global

Ethyl Alcohol with Deionized Water, 140 proof

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 gal.
4 x 1 gal.
55 gal.
Número de catálogo P111000140DM55
Precio (USD)
-
Cantidad:
55 gal.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición87°C to 90°C
CAS64-17-5
Nombre del producto químico o materialEthyl Alcohol
ColorIncolora
Fórmula molecularC2H6O
EnvasePoly drum
SinónimoEthanol, Ethyl Alcohol
Peso molecular (g/mol)46.07
Porcentaje de pureza70%
Cantidad55 gal.
Unit Size55 gal.