Ethyl Alcohol, 190 proof, GMP
Greenfield Global

Ethyl Alcohol, 190 proof, GMP

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Ansicht ändernbuttonViewtableView
Menge:
5 gal.
4 x 4 L
55 gal.
Katalognummer P111GMP190DM55UV
Preis (EUR)
-
Menge:
55 gal.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Siedepunkt87°C to 90°C
CAS64-17-5
Chemischer Name oder MaterialEthyl Alcohol
FarbeFarblos
SummenformelC2H6O
VerpackungPoly drum
SynonymEthanol, Ethyl Alcohol
GüteMulti-Kompendium
Molekulargewicht (g/mol)46.07
Reinheit (%)95%
Menge55 gal.
Unit Size55 gal.