Ethyl Alcohol with WFI, 35 proof
Greenfield Global

Ethyl Alcohol with WFI, 35 proof

This product is made with USP Grade Water for Injection (WFI).
Have Questions?
Catalog number P111WFI350DM55
Price (MXN)
-
This product is made with USP Grade Water for Injection (WFI).
This product is made with USP Grade Water for Injection (WFI). Greenfield Global USA Inc. is not certifying the finished product as injection grade. The suitability of the finished product for injection purposes is the sole responsibility of the end user. This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Boiling Point87°C to 90°C
CAS64-17-5
Chemical Name or MaterialEthyl Alcohol
ColorColorless
Molecular FormulaC2H6O
PackagingPoly drum
SynonymEthanol, Ethyl Alcohol
GradeNF/USP
Molecular Weight (g/mol)46.07
Percent Purity17.0–18.0%
Quantity200 L
Unit Size200 L