Ethyl Alcohol, 200 proof, Multi-Compendial, GMP
Ethyl Alcohol, 200 proof, Multi-Compendial, GMP
Greenfield Global

Ethyl Alcohol, 200 proof, Multi-Compendial, GMP

Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be usedMás información
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 gal.
4 x 4 L
4 x 1 gal.
55 gal.
Número de catálogo P111WORLD200DM55M
Precio (MXN)
-
Solicite un presupuesto
Cantidad:
55 gal.
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición87°C to 90°C
CAS64-17-5
Nombre del producto químico o materialEthyl Alcohol
ColorIncolora
Fórmula molecularC2H6O
EnvaseMetal drum
SinónimoEthanol, Ethyl Alcohol
GradoMulti-Compendial
Peso molecular (g/mol)46.07
Porcentaje de pureza100%
Cantidad55 gal.
Unit Size55 gal.