PFB-FDGlu (5-(Pentafluorobenzoylamino)Fluorescein Di-β-D-Glucopyranoside)
Invitrogen™

PFB-FDGlu (5-(Pentafluorobenzoylamino)Fluorescein Di-β-D-Glucopyranoside)

PFB-FDGlu is a cell-permeable substrate used for live cell imaging of GCase activity. It fluoresces green upon cleavage by lysosomal GCase, facilitating specific, single-cell analysis via high-content imaging, flow cytometry, and microscopy.
Have Questions?
Catalog NumberQuantity
P11947
also known as P-11947
5 mg
Catalog number P11947
also known as P-11947
Price (USD)
631.40
Special offer
Online exclusive
Ends: 31-Dec-2026
770.00
Save 138.60
Each
Quantity:
5 mg
Price (USD)
631.40
Special offer
Online exclusive
Ends: 31-Dec-2026
770.00
Save 138.60
Each
PFB-FDGlu (5-(Pentafluorobenzoylamino) Fluorescein Di-beta-D-Glucopyranoside) is a glycosidase activity probe for live cell imaging of lysosomal glucocerebrosidase (GCase) activity. This enzyme is vital for glycolipid metabolism, and its deficiency is linked to diseases like Gaucher’s and Parkinson's. PFB-FDGlu fluoresces green upon cleavage by GCase, allowing specific, single-cell analysis via flow cytometry, fluorescent microscopy, and high-content imaging.

PFB-FDGlu is cell-permeable and when loaded into cells, localizing to endosomal and lysosomal compartments while maintaining cell viability. Upon enzymatic cleavage by active GCase enzymes, it yields a green-fluorescent dye (PFB-F) with excitation/emission maxima of approximately 492/516 nm. This fluorescence can be detected using fluorescence microscopy, high-content imaging, and flow cytometry, allowing for specific, single-cell analysis of GCase activity.

Advantages of PFB-FDGlu for detecting GCase activity in lysosomes:

  • High specificity—utilizes a lysosomal GCase substrate for measurement of GCase activity, ensuring specific lysosomal staining (note that with long incubations the fluorescent PFB-F will migrate to the cytoplasm)
  • Live cell imaging—allows for the assessment of GCase activity in intact, living cells with minimal impact on cell viability, even with extended incubation
  • Single-cell analysis—enables specific, single-cell analysis with flow cytometry, high-content imaging, and microscopy to distinguish between different cell types
  • Bright fluorescence—yields a bright green fluorescent dye upon cleavage by GCase, enhancing detection sensitivity
  • Versatile protocol—can be combined with other phenotypic indicators, such as viability stains and antibodies, to identify distinct cell populations or as a selection criterion for cell sorting

GCase, encoded by the GBA1 gene, is a lysosomal hydrolase that converts glucosylceramide to glucose and ceramide. Mutations in GBA1 leading to GCase deficiency can cause lysosomal storage disorders. GCase deficiencies are implicated in several diseases, including Gaucher's disease and Parkinson's disease.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-Permeant
ColorGreen
Excitation/Emission492/526
For Use With (Equipment)Flow Cytometer, High Content Imaging Instrument, Fluorescent Microscope, Confocal Microscope
Label or DyeFluorescein
Product TypeSubstrate
Quantity5 mg
Shipping ConditionRoom Temperature
Storage RequirementsFreezer (-5°C to -30°C)
SubstrateGCase Substrate
Detection MethodFluorescence
FormPowder
Substrate PropertiesChemical Substrate
Substrate TypeBeta-Glucosidase Substrate
Target EnzymeBeta-Glucosidase
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).

Citations & References (16)

Citations & References
Abstract
Activation and Purification of ß-Glucocerebrosidase by Exploiting its Transporter LIMP-2 - Implications for Novel Treatment Strategies in Gaucher's and Parkinson's Disease.
Authors:Dobert JP,Bub S,Mächtel R,Januliene D,Steger L,Regensburger M,Wilfling S,Chen JX,Dejung M,Plötz S,Hehr U,Moeller A,Arnold P,Zunke F
Journal:Advanced science (Weinheim, Baden-Wurttemberg, Germany)
PubMed ID:38666485
Genetic variants of GBA1 can cause the lysosomal storage disorder Gaucher disease and are among the highest genetic risk factors for Parkinson's disease (PD). GBA1 encodes the lysosomal enzyme beta-glucocerebrosidase (GCase), which orchestrates the degradation of glucosylceramide (GluCer) in the lysosome. Recent studies have shown that GluCer accelerates α-synuclein aggregation, ... More
Late-onset Krabbe disease presenting as spastic paraplegia - implications of GCase and CTSB/D.
Authors:Mächtel R,Dobert JP,Hehr U,Weiss A,Kettwig M,Laugwitz L,Groeschel S,Schmidt M,Arnold P,Regensburger M,Zunke F
Journal:Annals of clinical and translational neurology
PubMed ID:38837642
Krabbe disease (KD) is a multisystem neurodegenerative disorder with severe disability and premature death, mostly with an infancy/childhood onset. In rare cases of late-onset phenotypes, symptoms are often milder and difficult to diagnose. We here present a translational approach combining diagnostic and biochemical analyses of a male patient with a ... More
The Parkinson's disease risk gene cathepsin B promotes fibrillar alpha-synuclein clearance, lysosomal function and glucocerebrosidase activity in dopaminergic neurons.
Authors:Jones-Tabah J,He K,Karpilovsky N,Senkevich K,Deyab G,Pietrantonio I,Goiran T,Cousineau Y,Nikanorova D,Goldsmith T,Del Cid Pellitero E,Chen CX,Luo W,You Z,Abdian N,Ahmad J,Ruskey JA,Asayesh F,Spiegelman D,Fahn S,Waters C,Monchi O,Dauvilliers Y,Dupré N,Miliukhina I,Timofeeva A,Emelyanov A,Pchelina S,Greenbaum L,Hassin-Baer S,Alcalay RN,Milnerwood A,Durcan TM,Gan-Or Z,Fon EA
Journal:Molecular neurodegeneration
PubMed ID:39587654
Variants in the CTSB gene encoding the lysosomal hydrolase cathepsin B (catB) are associated with increased risk of Parkinson's disease (PD). However, neither the specific CTSB variants driving these associations nor the functional pathways that link catB to PD pathogenesis have been characterized. CatB activity contributes to lysosomal protein degradation ... More
Dysregulation of mitochondria-lysosome contacts by GBA1 dysfunction in dopaminergic neuronal models of Parkinson's disease.
Authors:Kim S,Wong YC,Gao F,Krainc D
Journal:Nature communications
PubMed ID:33753743
Mitochondria-lysosome contacts are recently identified sites for mediating crosstalk between both organelles, but their role in normal and diseased human neurons remains unknown. In this study, we demonstrate that mitochondria-lysosome contacts can dynamically form in the soma, axons, and dendrites of human neurons, allowing for their bidirectional crosstalk. Parkinson's disease ... More
Development of screening strategies for the identification of paramylon-degrading enzymes.
Authors:Gissibl A,Care A,Sun A,Hobba G,Nevalainen H,Sunna A
Journal:Journal of industrial microbiology & biotechnology
PubMed ID:30806871
Enzymatic degradation of the β-1,3-glucan paramylon could enable the production of bioactive compounds for healthcare and renewable substrates for biofuels. However, few enzymes have been found to degrade paramylon efficiently and their enzymatic mechanisms remain poorly understood. Thus, the aim of this work was to find paramylon-degrading enzymes and ways ... More