Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Simplify field preservation with Thermo Scientific™ Chemically-Preserved Environmental Sample Containers and enjoy peace of mind as well as convenience. Using certified containers and chemicals reduces the risk of container and preservative contamination. Let us save you the time, tedium and chemical contact associated with diluting and aliquoting chemical preservatives to prepare environmental sample container sets. Choose from our standard offering of chemically-preserved containers, or let us custom-create container/preservative combos according to your personal analytical needs.
Diameter (English) Outer | 1.1 in. |
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 5 mL Methanol, tare weighed |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 0.5 mL 1:1 Hydrochloric Acid |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 0.25 mL 1:1 Hydrochloric Acid |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 0.2 mL 1:1 Hydrochloric Acid |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Amber |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 0.2 mL 1:1 Hydrochloric Acid |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Amber |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 0.5 mL 1:1 Hydrochloric Acid |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 10 mL Methanol, tare weighed |
Closure Size | 24-414 |
Unit Size | Each |
Diameter (English) Outer | 1.1 in. |
Color | Clear |
Diameter (Metric) Outer | 28 mm |
Material | Glass (Type 1) |
Volume (Metric) | 40 mL |
Height (English) | 3.75 in. |
Level | Certified |
No. per Case | 72 |
Volume (English) | 1 oz. |
Volume (Metric) Total | 40 mL |
Closure Type | Septa Cap |
Height (Metric) | 95 mm |
Thickness (Metric) | 3.17 mm |
Certifications/Compliance | Certified to meet the EPA Performance Based Specifications for volatile organic compounds; packaged |
Type | Environmental Sample Container |
Preservative | 5 mL Sodium Bisulfate, Stir Bar, tare weighed |
Closure Size | 24-414 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Volume (Metric) | Color | Preservative | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
P140-40CEPPTTW | Each | 40 mL | Clear | 5 mL Methanol, tare weighed | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP140-40CEP.5HA | Each | 40 mL | Clear | 0.5 mL 1:1 Hydrochloric Acid | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP140-40CEP.25HA | Each | 40 mL | Clear | 0.25 mL 1:1 Hydrochloric Acid | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP140-40CEP.2HA | Each | 40 mL | Clear | 0.2 mL 1:1 Hydrochloric Acid | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP141-40AEP.2HA | Each | 40 mL | Amber | 0.2 mL 1:1 Hydrochloric Acid | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP141-40AEP.5HA | Each | 40 mL | Amber | 0.5 mL 1:1 Hydrochloric Acid | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP140-40CEPPTTW | Each | 40 mL | Clear | 10 mL Methanol, tare weighed | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
PP140-40CEPSBTB | Each | 40 mL | Clear | 5 mL Sodium Bisulfate, Stir Bar, tare weighed | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Ordering Information:
Custom preserved containers available by special order:
Products available only in the US
Includes:
Certified sample containers dosed with chemical preservative; certificate of analysis for container and preservative, individually barcoded containers traceable to certs, sample analysis labels unattached for optional use, hazardous material compliant packaging. MSDS available upon request.