P2-CST Multiple Gradient Gel Caster Comb, 25 teeth, 1.5mm tooth thickness
P2-CST Multiple Gradient Gel Caster Comb, 25 teeth, 1.5mm tooth thickness
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
Have Questions?
Catalog NumberTeeth Count
P2-25D25
Catalog number P2-25D
Price (USD)
76.95
Online exclusive
85.50
Save 8.55
Each
Teeth Count:
25
Price (USD)
76.95
Online exclusive
85.50
Save 8.55
Each
Cast one to five gels simultaneously with the Fisherbrand™ P2-CST Multiple Gradient Gel Caster, one of the simplest devices to cast gradient gels.
  • With an external gradient maker, gels can be cast with ease and great reproducibility
  • The design and format of this unit will provide years of service in your lab
Specifications
DescriptionComb, 25 teeth; Tooth thickness, 1.5mm
Teeth Count25
Thickness (Metric) Teeth1.5 mm
For Use With (Equipment)Fisherbrand P2-CST System
TypeComb
Unit SizeEach