Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit
Invitrogen™

Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit

Pro-Q™ Emerald 300 Lipopolysaccharide Gel Stain Kit
Have Questions?
Catalog NumberQuantity
P204951 kit
Catalog number P20495
Price (USD)
390.00
Each
Quantity:
1 kit
Price (USD)
390.00
Each

Molecular Probes' proprietary Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation, making these stains at least 50-fold more sensitive than standard fuchsin staining.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Detection LocationIn-Blot Detection, In-Gel Detection
Detection MethodFluorescence
Quantity1 kit
Shelf Life6 Months
Target MoleculeLipopolysaccharide
Label or DyePro-Q Emerald 300
Product LinePRO-Q
Product TypeLipopolysaccharide Stain Kit
Unit SizeEach

Frequently asked questions (FAQs)

I stained my Invitrogen NuPAGE Bis-Tris gel with Pro-Q Emerald Glycoprotein Gel Stain and observe a very high background. What is causing this?

Pro-Q Emerald Glycoprotein Gel Stain may show high background staining in Invitrogen NuPAGE Bis-Tris and Tris-Acetate gels, especially in combination with MES running buffer or in gels that are nearing their expiration date. The gel background increases with acrylamide density and gradient gels will show a gradual increase in background from the top to the bottom of the gel corresponding to the acrylamide gradient. Increasing the number of washes or modifying incubation times will not help to reduce this background. Better results will be obtained with Tris-Glycine or Tricine gels. If you wish to continue using Pro-Q Emerald stain with Invitrogen NuPAGE Bis-Tris gels, we recommend using recently purchased gels and MOPS running buffer. Glycoproteins will still be detected in gels with high background, but with reduced sensitivity.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am observing speckling in my gel stained with Pro-Q Emerald 300/488 Glycoprotein Gel Stain, especially near the edges. What is causing this?

Speckling of Pro-Q Emerald dye, especially with Pro-Q Emerald 300 stain, can occur as the Pro-Q Emerald dye ages, due to self-aggregation of the dye over time. Speckles can also form due to dye binding to contaminants from the staining container, solutions, or particles from the air or gloves. Non-dye speckles can also show up in the image from auto-fluorescent particles of dust, hair, glove powder, or clothing lint that falls on the gel or surface of the glass imaging plate. The better the imager is at focusing on surface features of the gel, the more speckles that are going to be visible.

To minimize the formation of speckles and other background debris, follow clean laboratory practices, use ultrapure water of >18 megohm-cm resistance to prepare solutions, rinse gloves in water to remove powder residue before touching gels, use lint-free wipes and wear a lab coat or avoid wearing clothing that generates a lot of lint, always rinse the staining container with ethanol and wipe out any residual dye before staining another gel, and always rinse and wipe down the glass imaging surface with ethanol and water before placing your gel down. Once speckles have been deposited on the gel, it is not possible to wash them off. When analyzing amounts of glycoprotein near the limit of detection, we advise running samples in the middle lanes of the gel.

Speckles will show up as sharp, tall spikes on 3D renditions of gel images. These spikes look distinct from 3D renditions of protein spots or bands. Some image analysis software packages have de-speckling algorithms that can easily identify and remove this type of pixelated noise.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

I am observing poor specific signal in my gel stained with Pro-Q Emerald 300/488 Glycoprotein Gel Stain, or also observe a weak total protein staining pattern when I switch to a different excitation or emission filter. What is causing this?

Many total protein stains including SYPRO Ruby Gel Stain and Coomassie Blue stain will quench the Pro-Q Emerald signal. If you are staining your gels or blots with Pro-Q Emerald stain in containers that have previously been used for a total protein stain, you may be contaminating your gel with residue left on the staining dish from the total protein stain. Either use new containers, such as plastic weigh boats, designated containers for each stain, or rinse the container well in ethanol and wipe out any residual residue with a Kimwipe tissue.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

My gel stained with Pro-Q Emerald 300/488 Glycoprotein Gel Stain is showing poor staining of my glycoproteins and the glycosylated CandyCane molecular weight standard. What is causing this and how can I improve the signal?

- Poor staining can be caused by the presence of primary amines, such as from Tris or glycine, that will also react with the aldehyde/ketone groups generated by periodate oxidation. This effectively caps the reactive groups, leaving no reactive sites for Pro-Q Emerald dye to bind. To remove any amine contamination, increase the volume or number of incubations in fresh fixative and then increase the volume or number of washes in wash buffer.
- Poor staining can also be due to inadequate removal of the periodic acid oxidation solution. Increase the volume or number of washes after the oxidation step to ensure adequate removal of periodic acid.
- Poor staining can also be due to inadequate oxidation of glycoprotein sugars. Increase the volume of periodic acid oxidation solution.

Note: Do not increase the number or incubation time of the periodic acid oxidation in excess of 30 minutes for small-format gels. Excessive periodic acid oxidation could result in increased staining of non-glycosylated proteins. In general, large format, unusually thick or very high percent acrylamide gels may require additional incubations or wash steps for optimal signal and sensitivity of staining with Pro-Q Emerald Glycoprotein Gel Stain.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

My gel stained with Pro-Q Emerald 300/488 Glycoprotein Gel Stain is showing some staining of non-glycosylated proteins, but only the four glycoprotein bands in the CandyCane marker lane. What is causing this?

If the CandyCane standard and test glycoproteins are staining correctly, then the kit is performing well. Some very highly abundant proteins, such as albumin in serum and plasma, may stain lightly. The Pro-Q Emerald dye is covalently attached to sugar residues, so more post-staining washes will help to remove any non-covalently bound dye. Non-specific staining due to high abundance can be determined by post-staining with a total protein stain, such as SYPRO Ruby Protein Gel Stain. That being said, some proteins are actually heavily oxidized in the native state, and this carbonylation will be picked up by the Pro-Q Emerald reagent. Carbonylation of amino acids can be distinguished from glycosylation by performing the Pro-Q Emerald staining without the oxidation step. Under these conditions, the CandyCane marker bands will not be stained, but the carbonylated proteins will.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Citations & References (12)

Citations & References
Abstract
The heat sensitivity of cytokine-inducing effect of lipopolysaccharide.
Authors:Gao B, Wang Y, Tsan MF,
Journal:J Leukoc Biol
PubMed ID:16720829
Heat inactivation by boiling has been widely used as a criterion to determine whether the observed effects of a protein preparation are a result of lipopolysaccharide (LPS) contamination. However, the heat sensitivity of LPS cytokine-inducing activity has not been characterized. In the current study, we demonstrated that the endotoxin activity, ... More
Bacterial cell wall synthesis gene uppP is required for burkholderia colonization of the Stinkbug Gut.
Authors:Kim JK, Lee HJ, Kikuchi Y, Kitagawa W, Nikoh N, Fukatsu T, Lee BL,
Journal:
PubMed ID:23747704
'To establish a host-bacterium symbiotic association, a number of factors involved in symbiosis must operate in a coordinated manner. In insects, bacterial factors for symbiosis have been poorly characterized at the molecular and biochemical levels, since many symbionts have not yet been cultured or are as yet genetically intractable. Recently, ... More
A cyanobacterial lipopolysaccharide antagonist inhibits cytokine production induced by Neisseria meningitidis in a human whole-blood model of septicemia.
Authors:Jemmett K, Macagno A, Molteni M, Heckels JE, Rossetti C, Christodoulides M,
Journal:Infect Immun
PubMed ID:18443097
'Septicemia caused by Neisseria meningitidis is characterized by increasing levels of meningococcal lipopolysaccharide (Nm-LPS) and cytokine production in the blood. We have used an in vitro human whole-blood model of meningococcal septicemia to investigate the potential of CyP, a selective Toll-like receptor 4 (TLR4)-MD-2 antagonist derived from the cyanobacterium Oscillatoria ... More
Colonial morphology of Burkholderia cepacia complex genomovar III: implications in exopolysaccharide production, pilus expression, and persistence in the mouse.
Authors:Chung JW, Altman E, Beveridge TJ, Speert DP
Journal:Infect Immun
PubMed ID:12540572
'The purpose of this study was to determine the role of colonial morphology of Burkholderia cepacia complex (BCC) organisms in pathogenicity in a mouse model of pulmonary infection. BCC strain C1394 was rapidly cleared by leukopenic mice after intranasal challenge, whereas a spontaneous variant (C1394mp2) that was indistinguishable from the ... More
The PmrA/PmrB regulatory system controls the expression of the wzzfepE gene involved in the O-antigen synthesis of Salmonella enterica serovar Typhimurium.
Authors:Pescaretti Mde L, López FE, Morero RD, Delgado MA,
Journal:Microbiology
PubMed ID:21719537
'The degree of polymerization of O-antigen from Salmonella enterica serovar Typhimurium is controlled by the products of the wzz(s)(t) and wzz(fepE) genes. In the present study we investigated the role of the PmrA/PmrB regulatory system in wzz(fepE) transcription. We report that the direct binding of the PmrA regulator to a ... More