Isopropanol, GMP, Multi-Compendial
Greenfield Global

Isopropanol, GMP, Multi-Compendial

This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
4 x 1 gal.
55 gal.
Número de catálogo P211USPNFCSGL
Precio (MXN)
-
Cantidad:
4 x 1 gal.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
This product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición83°C
CAS67-63-0
Nombre del producto químico o materialIsopropyl Alcohol
ColorIncolora
Fórmula molecularC3H8O
EnvaseCase
SinónimoIsopropanol, Isopropyl Alcohol, IPA
GradoUSP
Peso molecular (g/mol)18.02
Porcentaje de pureza68.0–72.0%
Cantidad4 x 1 gal.
Unit Size4x1 gal.