Related applications: Cell Viability, Proliferation & Function | Pharma & Biopharma | Protein Assays and Analysis | Target-Based ADME⁄Tox Assays
Error loading your content!
Catalog number |
Select a plan
Unit size
|
Price ({{currency}}) | Availability | Qty | |
---|---|---|---|---|---|
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} |
|
— |
Detection Method: | Fluorescent |
---|---|
Product Line: | VIVID® |
Product Size: | 1 mg |
Substrate: | OOMR |
Substrate Properties: | Chemical Substrate |
Substrate Type: | Cytochrome P450 Substrate |
Shipping Condition: | Approved for shipment on Wet or Dry Ice |
Catalog number: | P2859 |
Size | 300 assays |
Price | USD 394.00 |
Add to favorites | |
Catalog number: | P2378 |
Size | 0.5 nmol |
Price | USD 193.00 |
Add to favorites | |
Catalog number: | P2857 |
Size | 300 assays |
Price | USD 394.00 |
Add to favorites | |