Acetonitrile, GMP
Greenfield Global

Acetonitrile, GMP

Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Número de catálogo P300000GMPDM55M
Precio (CLP)
-
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.

Para su uso en investigación o procesos de fabricación posteriores. No apto para uso diagnóstico ni para la administración directa en seres humanos ni en animales.

Especificaciones
Punto de ebullición178.9°F (81.6°C) at 101.325 kPa
CAS75-05-8
Nombre del producto químico o materialAcetonitrile
ColorClear
Fórmula molecularC2H3N
EnvaseMetal drum
SinónimoEthanenitrile, Methyl Cyanide, Cyanomethane
GradoUSP
Peso molecular (g/mol)41.05
Porcentaje de pureza100%
Cantidad55 gal.
Unit Size55 gal.