Pacific Orange™ and Pacific Blue™ Antibody Labeling Kits
Invitrogen™

Pacific Orange™ and Pacific Blue™ Antibody Labeling Kits

Antibody Labeling Kits specifically bind small amounts of antibody with violet light-excitable Pacific Orange or Pacific Blue dyes. Conveniently label up to 100 μg of antibody per reaction, with 5 reactions per kit.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberExcitation/EmissionColor
P30013410/455 nmPacific Blue
P30014405/551 nmPacific Orange
Catalog number P30013
Price (USD)
672.00
Each
Excitation/Emission:
410/455 nm
Color:
Pacific Blue
Request bulk or custom format
Price (USD)
672.00
Each
Pacific Orange and Pacific Blue antibody labeling kits enable attachment of a violet light-excitable fluorescent label on antibodies (up to 100 μg per reaction). Each kit contains enough dye for five reactions.

A primary antibody directly labeled with a fluorophore often produces lower background fluorescence and less nonspecific binding. Primary antibodies of the same isotype or derived from the same species can be multiplexed if they are directly labeled with compatible fluorophores. Using Pacific Blue and Pacific Orange dyes simultaneously allows you to excite both dyes at the same wavelength while their emissions are differentially distinguished.

Pacific Orange and Pacific Blue antibody labeling kits allow the labeling of small amounts of antibody with a violet light-excitable dye (up to 100 μg of antibody per reaction). Small amounts of other proteins (>40 kDa) can also be labeled using these kits.

Features of these antibody labeling kits
• Pacific Blue excitation and emission maxima: 410/455 nm
• Pacific Orange excitation and emission maxima: 405/551 nm
• Labeled proteins typically ready to use in 90 min. (∼15 min. hands-on time)
• Optimized for small-scale labeling of any protein >40 kDa
• Stabilizing proteins should be removed from the sample before labeling
• Purified using convenient spin filters
• Includes detailed instructions for determining degree of labeling (DOL)

These kits contain everything you need to perform five separate labeling reactions as well as to purify the resulting conjugates. Conjugates are ideal for multiple applications, including flow cytometry, fluorescent microscopy, immunohistochemistry, primary detection, ELISA, immunocytochemistry, indirect FISH, and more. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
ColorPacific Blue
Detection MethodFluorescence
Excitation/Emission410/455 nm
Label TypePacific Dyes
Labeling Scale100 μg
Product LinePacific Blue
Product TypeAntibody Labeling Kit
Quantity5 Reactions kit
Shipping ConditionRoom Temperature
Chemical ReactivityAmine
Labeling TargetAntibodies, Proteins
Label or DyePacific Blue
SolubilityDMSO (Dimethylsulfoxide)
Unit SizeEach
Contents & Storage
Contains 5 vials of Pacific Blue™ reactive dye, 1 vial sodium bicarbonate (84 mg), purification resin (10 mL), 5 spin columns, and and 5 collection tubes.

Store at 2-8°C, protected from light. Component A may be stored from -20°C to 8°C.

Frequently asked questions (FAQs)

What formulation of antibody should I use for conjugation for small animal in vivo imaging?

To allow for good reaction kinetics, antibodies should be in PBS buffer at a concentration of 0.5-3.0 mg/ml. The antibody must be free of preservatives (azide etc.), amine containing buffers and carrier proteins such as BSA.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is degree of labeling (DOL)?

Degree of labeling (DOL) describes the number of fluorophores per antibody. For in vivo labeling experiments, the DOL is restricted to a narrow range because it has significant consequences for the biodistribution and clearance of the probe. For example, for in vivo imaging, we have determined that the DOL range for the far-red Alexa Fluor dyes is 1.5 to 3 molecules per antibody for optimal optical in vivo imaging.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (1)

Citations & References
Abstract
PLC-gamma2 is essential for formation and maintenance of memory B cells.
Authors:Hikida M, Casola S, Takahashi N, Kaji T, Takemori T, Rajewsky K, Kurosaki T,
Journal:J Exp Med
PubMed ID:19273623
Resting antigen-experienced memory B cells are thought to be responsible for the more rapid and robust antibody responses after antigen reencounter, which are the hallmark of memory humoral responses. The molecular basis for the development and survival of memory B cells remains largely unknown. We report that phospholipase C (PLC) ... More