BlockAid™ Universal Blocking Solution—ready-to-use for beautiful fluorescent imagery. Learn more about this product.

Goat anti-Mouse IgG (H+L) Molecular Probes™ Secondary Antibody, Pacific Blue™ conjugate

Catalog number: P31582

Invitrogen™  Related applications: Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Wells: 12 wells
Adapters: No Adapters
Capacity: Up to 45 µL per well
Gel Size: Mini (8 cm x 8 cm)
Gel Type: Tris-Glycine Gel
Quantity: 10 gels
Thickness: 1.0 mm
Well Format: 1D Well (Wedge-Shaped)
Product Size: 10 gels (1 box)
Gel Percentage: 14 %
Separation Range: 15 kDa to 110 kDa
Mode of Separation: Molecular Weight
Sample Loading Volume: 30 µL (recommended)
For Use With (Equipment): Mini Gel Tank
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C) and protect from light.

Documents

Manuals & protocols

Recommended products