Methanol absolute, GMP, Multi-Compendial
Greenfield Global

Methanol absolute, GMP, Multi-Compendial

Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
4 x 4 L
55 gal.
Número de catálogo P339WORLDCS4L
Precio (USD)
-
Cantidad:
4 x 4 L
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Especificaciones
Punto de ebullición147.2°F (64°C) at 101.325 kPa
CAS67-56-1
Nombre del producto químico o materialMethanol
ColorIncolora
Fórmula molecularCH4O
EnvaseCase
SinónimoMethyl alcohol
GradoMulti Compendial
Peso molecular (g/mol)32.04
Porcentaje de pureza≥99.8%
Cantidad4 x 4 L
Unit Size4 x 4 L