Methanol absolute, GMP, Multi-Compendial
Greenfield Global

Methanol absolute, GMP, Multi-Compendial

Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Have Questions?
Modifier l'affichagebuttonViewtableView
Quantité:
4 x 4 L
55 gal.
Référence P339WORLDDM55UV
Prix (EUR)
-
Quantité:
55 gal.
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes.
Manufactured in compliance with cGMP, this product is for further commercial manufacturing, laboratory or research use, and may be used as an excipient or a process solvent for pharmaceutical purposes. It is not intended for use as an active ingredient in drug manufacturing, nor as a medical device or disinfectant. Appropriate/legal use of this product is the responsibility of the user.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Spécifications
Point d’ébullition147.2°F (64°C) at 101.325 kPa
CAS67-56-1
Nom chimique ou matériauMethanol
CouleurIncolore
Formule moléculaireCH4O
ConditionnementUV drum
SynonymeMethyl alcohol
QualitéMulti Compendial
Poids moléculaire (g/mol)32.04
Pourcentage de pureté≥99.8%
Quantité55 gal.
Unit Size55 gal.