pHrodo™ Red and Green AM Intracellular pH Indicator Dyes
pHrodo™ Red and Green AM Intracellular pH Indicator Dyes
pHrodo™ Red and Green AM Intracellular pH Indicator Dyes
pHrodo™ Red and Green AM Intracellular pH Indicator Dyes
pHrodo™ Red and Green AM Intracellular pH Indicator Dyes
Invitrogen™

pHrodo™ Red and Green AM Intracellular pH Indicator Dyes

Quantify intracellular pH with pHrodo Red and Green AM intracellular pH indicator dyes, which permeate cell membranes and fluoresce in acidic cytosolic environments. Both the pHrodo Red and Green AM indicator dyes are compatible with HCS, fluorescence microplate, flow cytometry, and HTS platforms.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
P35373Green
P35372Orange
Catalog number P35373
Price (USD)
523.16
Special offer
Online exclusive
Ends: 31-Dec-2026
638.00
Save 114.84
Each
Color:
Green
Price (USD)
523.16
Special offer
Online exclusive
Ends: 31-Dec-2026
638.00
Save 114.84
Each
Measure and quantify intracellular pH with pHrodo Red and Green AM indicator dyes, which are fluorogenic probes that traverse the cell membrane and remain within the intracellular space upon cleavage by nonspecific esterases. pHrodo Red and Green AM indicator dyes become increasingly fluorescent as the pH drops and can be used to quantify cytosolic pH in the range of 4–9. These indicator dyes can also be used in multiplex experiments within HCS, flow cytometry, microplate fluorometry, and HCS applications.
pHrodo Red (Cat. No. P35372) and Green (Cat. No. P35373) AM intracellular pH indicators are fluorogenic probes used to measure intracellular pH in live cells. They are weakly fluorescent at neutral pH but become increasingly fluorescent as the pH decreases. These reagents can be used to quantify cellular cytosolic pH in the range of 4–9 with a pKa of ∼6.5 and with excitation/emission spectra of 560/585 nm (pHrodo Red) or 509/533 nm (pHrodo Green).

Both pHrodo Red and pHrodo Green pH indicator dyes are photostable and appropriate for use in multiplexing experiments. pHrodo Red easily multiplexes with a wide variety of blue, green, and far red dyes and reporters such as GFP, Fuo-4, calcein, NucBlue, CellEvent Caspase 3/7 green, Mitosox Green, and Mitotracker Deep Red, among many others. pHrodo Green easily multiplexes with a wide variety of blue, red, and far red dyes reporters such as Mitosox Red, CellEvent Caspase 3/7 Red, NucBlue, RFPs, and Mitotracker Deep Red, among many others.

The pHrodo Red and Green dyes have been modified with AM ester groups, resulting in an uncharged molecule that can permeate cell membranes. Once inside the cell, the lipophilic blocking groups of the dyes are cleaved by nonspecific esterases, resulting in compounds that are retained within the intracellular space. The fluorescence intensity of the probe then becomes an indicator of intracellular pH. Subsequent use of the Intracellular pH Calibration Buffer Kit (Cat. No. P35379) allows this intracellular pH to be quantified.

pHrodo Red and Green AM intracellular pH indicator dyes are compatible with various platforms, such as traditional fluorescence microscopy, high content screening (HCS), flow cytometry, and microplate-based fluorimetry or high throughput screening (HTS). These reagents are also compatible with widefield or confocal fluorescence microscopy, flow cytometry, fluorescence plate readers, and high content instruments.

For Research Use Only. Not for human or animal therapeutic or diagnostic use.
Specifications
DescriptionpHrodo™ Green AM Intracellular pH Indicator
Detection MethodFluorescence
Dye TypepH Indicator
Emission WavelengthVisible
FormLiquid
Quantity50 μL
SolubilityDMSO (Dimethylsulfoxide)
Sub Cellular LocalizationCytoplasm
ColorGreen
For Use With (Application)Cell Analysis
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer, Fluorometer, High Content Analysis Instrument
Product LinepHrodo
Product TypepH Indicator
Unit SizeEach
Contents & Storage
Store at 2–8°C, desiccate, and protect from light.

Citations & References (11)

Citations & References
Abstract
Macrophage Metabolism of Apoptotic Cell-Derived Arginine Promotes Continual Efferocytosis and Resolution of Injury.
Authors:
Journal:Cell Metab
PubMed ID:32004476
Carbonic anhydrase 12 mutation modulates membrane stability and volume regulation of aquaporin 5.
Authors:Hwang S, Kang JY, Kim MJ, Shin DM, Hong JH
Journal:J Enzyme Inhib Med Chem
PubMed ID:30451023
'Patients carrying the carbonic anhydrase12 E143K mutation showed the dry mouth phenotype. The mechanism underlying the modulation of aquaporin 5 and function in the salivary glands by carbonic anhydrase12 remains unknown. In this study, we identified the mislocalised aquaporin 5 in the salivary glands carrying the E143K. The intracellular pH ... More
Recombinant annexin A6 promotes membrane repair and protects against muscle injury.
Authors:
Journal:J Clin Invest
PubMed ID:31545299
Role of amoebae for survival and recovery of 'non-culturable' Helicobacter pylori cells in aquatic environments.
Authors:
Journal:FEMS Microbiol Ecol
PubMed ID:32897313
NAD+ consumption by PARP1 in response to DNA damage triggers metabolic shift critical for damaged cell survival.
Authors:
Journal:Mol Biol Cell
PubMed ID:31390283