pHrodo™ Antibody Labeling Kits
Invitrogen™
pHrodo™ Antibody Labeling Kits
Perform antibody internalization, endocytosis, and phagocytosis experiments with confidence and ease using the updated pH-sensitive, brightly fluorescing dyes and ready-to-use spin columns in the pHrodo Red and Green Antibody Labeling Kits.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidadExcitación/emisiónEtiqueta o tinte
P360151 Kit505/525 nmpHrodo Green
P353553 x 100 μg640/655 nmpHrodo Deep Red
P353563 x 1 mg640/655 nmpHrodo Deep Red
P360203 x 100 μg560/585 nmpHrodo Red
P360213 x 1 mg560/585 nmpHrodo Red
P360223 x 100 μg505/525 nmpHrodo Green
P360233 x 1 mg505/525 nmpHrodo Green
P360141 Kit560/585pHrodo Red
Número de catálogo P36015
Precio (MXN)
-
Cantidad:
1 Kit
Excitación/emisión:
505/525 nm
Etiqueta o tinte:
pHrodo Green
Elevate biopharmaceutical research with pHrodo Antibody Labeling kits, designed to characterize antibody internalization, a critical step during antibody-drug conjugate research and development. Kits provide a simple, detailed protocol and advanced purification columns to easily label and purify antibodies and other large proteins. Amine-reactive pHrodo dyes covalently bind to antibodies and other proteins to produce bright, soluble conjugates whose fluorescence increases in acidic organelles including endosomes and lysosomes. Antibody internalization can be evaluated using flow cytometry, fluorescence microscopy, and high-content screening.
The pHrodo Red Antibody Labeling Kit and pHrodo Green Antibody Labeling Kit provide the reagents needed to easily label three purified protein or antibody samples with pHrodo Red or pHrodo Green dye. These kits contain amine-reactive, pH-sensitive pHrodo Red or Green STP ester dye that covalently binds to free lysines anywhere they occur in an antibody or protein.
The dyes included in this kit are improved versions of pHrodo Red and pHrodo Green dyes. They are optimized for the creation of bright and soluble bioconjugates that are used in the study of antibody internalization, endocytosis, phagocytosis, and for development of antibody-drug conjugates. These dyes are also more soluble than the original pHrodo Red and pHrodo Green dyes, making them useful for the labeling of antibodies that may otherwise precipitate out of solution during conjugation. pHrodo Red and pHrodo Green dyes are non-fluorescent outside the cell, but fluoresce bright red or bright green as the pH of their surroundings becomes more acidic, such as those found inside of phagosomes and endosomes.
The pHrodo Red and Green Microscale Labeling Kits offer the following benefits:
• Convenience—Labeling typically takes one hour, with less than 30 minutes of hands-on time
• Versatility—Labelling can be performed on 12-150 kDa molecular weight proteins
• Customization—Create your own probes for studying antibody internalization, phagocytosis, and endocytosis
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
ColorVerde
Excitación/emisión505/525 nm
Para utilizar con (aplicación)Análisis celular
Para utilizar con (equipo)Analizador de alto contenido
Línea de productospHrodo
Tipo de productoKit de etiquetado de proteínas
Cantidad1 Kit
Condiciones de envíoTemperatura ambiente
Suficiente para3 Labels
Etiqueta o tintepHrodo Green
Unit SizeEach
Contenido y almacenamiento
Almacenar en refrigerador de 2 °C a 8 °C, desecar, proteger de la luz y no congelar.