ProLong™ Live Antifade Reagent, for live cell imaging
ProLong™ Live Antifade Reagent, for live cell imaging
ProLong™ Live Antifade Reagent, for live cell imaging
Invitrogen™

ProLong™ Live Antifade Reagent, for live cell imaging

Prolong Live Antifade Reagent helps prevent the loss of fluorescent signal (photobleaching) during live cell imaging.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
P369751 x 1 mL
P369745 x 1 mL
Catalog number P36975
Price (USD)
69.91
Special offer
Online exclusive
Ends: 31-Dec-2026
85.25
Save 15.34
Each
Quantity:
1 x 1 mL
Price (USD)
69.91
Special offer
Online exclusive
Ends: 31-Dec-2026
85.25
Save 15.34
Each

ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through use of enzymes from the plasma membrane of E. coli. These enzymes metabolize the elements that can cause photobleaching. When dyes are illuminated, they can degrade which reduces the sample's fluorescent intensity and leads to the creation of free radical singlet oxygen which degrades neighboring dye molecules. ProLong Live Antifade Reagent metabolizes this element, increasing the signal intensity and duration while keeping the background signal low. ProLong Live reagent has little-to-no effect on cellular viability, proliferation, or other cellular functions in experiments up to 48 hours. ProLong Live reagent provides photobleach protection in live cell experiments for fluorophores such as GFP, RFP, Hoechst 33342, MitoTracker™, LysoTracker™, and CellTracker™dyes. ProLong Live Antifade Reagent can be added to any cell culture media or buffer that is suitable for fluorescent imaging.

ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through use of enzymes from the plasma membrane of E. coli. These enzymes metabolize the elements that can cause photobleaching. When dyes are illuminated, they can degrade which reduces the sample’s fluorescent intensity and leads to the creation of free radical singlet oxygen which degrades neighboring dye molecules. ProLong Live Antifade Reagent metabolizes this element, increasing the signal intensity and duration while keeping the background signal low. ProLong Live reagent has little-to-no effect on cellular viability, proliferation, or other cellular functions in experiments up to 48 hours. ProLong Live reagent provides photobleach protection in live cell experiments for fluorophores such as GFP, RFP, Hoechst 33342, MitoTracker™, LysoTracker™, and CellTracker™dyes. ProLong Live Antifade Reagent can be added to any cell culture media or buffer that is suitable for fluorescent imaging.

Specifications
Quantity1 x 1 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product LineProLong
Product TypeLive Antifade Reagent
Reagent TypeCell Imaging Reagent
Volume (Metric)1 mL
Unit SizeEach
Contents & Storage
When stored at -20°C product is stable for at least 6 months, with up to 4 freeze-thaw cycles. When stored at 2–8°C product should be used within 30 days (a precipitant may form at this temperature).

Frequently asked questions (FAQs)

After using ProLong Live Antifade Reagent on live samples and then fixing and permeabilizing, may the samples be mounted in ProLong Gold or Diamond Anifade Mountant?

Yes, this is possible.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After using ProLong Live Antifade Reagent on my live sample, may I fix and permeabilize the sample?

Yes. We recommend washing off the ProLong Live Antifade Reagent with pre-warmed buffer or media and proceeding to your standard fixation and permeabilization steps.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

May I use ProLong Live Antifade Reagent on a fixed/permeabilized sample?

This is possible for immediate viewing, but the sample cannot be archived with ProLong Live Antifade Reagent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to mount my dye-labeled cells in an antifade mounting medium to keep the dyes from photobleaching. Which mounting medium do you recommend?

As dyes are illuminated for imaging, they will fade, or “photobleach”, leading to unwanted dimming and lower detection efficiency over time. An antifade mounting medium can greatly reduce photobleaching. If you wish to label live cells, use of ProLong Live Antifade Reagent is helpful. If you wish to mount fixed cells after labeling, and then image immediately and then discard, SlowFade Diamond Antifade Mountant stays liquid but has good refractive index. If you wish to mount your sample and then archive the slides, ProLong Diamond Antifade Mountant will harden to a better refractive index and allow for archiving of the sample for up to several weeks, or even months. Unlike other antifade mounting media, these work well with fluorescent proteins for immediate viewing (archiving fluorescent proteins is not possible), and they are packaged with or without DAPI. More information on these can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/fluorescence-microscopy-and-immunofluorescence-if/mounting-medium-antifades/prolong-gold-antifade.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

My fluorescent dye signal is fading as I image it. What can I do to stop this?

All fluorescent dyes will fade, or “photobleach”, to at least some extent when exposed to strong light at the wavelengths they absorb. Here are some causes for photobleaching and ways to fix the problem:

1) Cause of photobleacing - Generation of free radicals and singlet oxygen
Remedy - i) Use an antifade reagent, which has antioxidants and free radical scavengers:

ii) For live-cell imaging of fluorescent dyes and proteins, we recommend ProLong Live Antifade Reagent which can be added to the cell media or buffer. ProLong Live Antifade Reagent can significantly increase the stability over time for reagents as well as fluorescent proteins, like GFP, without affecting cell health, for up to 24 hours.

iii) For immediate analysis and short-term storage of fixed samples, we recommend SlowFade Diamond Antifade Mountant (it is a liquid mountant and can be used for immediate viewing and then disposal of the sample within a day).

iv) For long-term analysis of Alexa Fluor dyes in fixed samples, we recommend a mountant that hardens, such as ProLong Diamond Antifade Mountant. The harrdening of the mountant also slows diffusion of free radicals).

v) For long-term analysis of all dyes and fluorescent proteins in fixed samples, we recommend ProLong Diamond Antifade Mountant, suitable for archiving slides.

2) Cause of photobleaching - Dye is particularly sensitive to structural modification upon exposure to light.
Remedy - i) Choose a more photostable dye, such as many of our Alexa Fluor dyes.

3) Cause of photobleaching - Intense Illlumination
Remedy - i) Reduce light exposure, for example by reducing laser power or using neutral density filters.

ii) Minimize the viewing time of labeled sample, and close shutter when not viewing.

iii) Use an objective with a lower numerical aperture, such as a lower-power objective.

You can find more information on choosing an antifade reagent on the link below http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/fluorescence-microscopy-and-immunofluorescence-if/mounting-medium-antifades.html.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (13)

Citations & References
Abstract
Targeting host mitochondria: A role for the Trypanosoma cruzi amastigote flagellum.
Authors:Lentini G, Dos Santos Pacheco N, Burleigh BA,
Journal:Cell Microbiol
PubMed ID:29119655
'Trypanosoma cruzi is the kinetoplastid protozoan parasite that causes human Chagas disease, a chronic disease with complex outcomes including severe cardiomyopathy and sudden death. In mammalian hosts, T. cruzi colonises a wide range of tissues and cell types where it replicates within the host cell cytoplasm. Like all intracellular pathogens, T. cruzi ... More
Methamphetamine Augments Concurrent Astrocyte Mitochondrial Stress, Oxidative Burden, and Antioxidant Capacity: Tipping the Balance in HIV-Associated Neurodegeneration.
Authors:Borgmann K, Ghorpade A,
Journal:Neurotox Res
PubMed ID:28993979
'Methamphetamine (METH) use, with and without human immunodeficiency virus (HIV)-1 comorbidity, exacerbates neurocognitive decline. Oxidative stress is a probable neurotoxic mechanism during HIV-1 central nervous system infection and METH abuse, as viral proteins, antiretroviral therapy and METH have each been shown to induce mitochondrial dysfunction. However, the mechanisms regulating mitochondrial ... More
Increased Fc?RIIB dominance contributes to the emergence of resistance to therapeutic antibodies in chronic lymphocytic leukaemia patients.
Authors:Burgess M, Mapp S, Mazzieri R, Cheung C, Chambers L, Mattarollo SR, Mollee P, Gill D, Saunders NA
Journal:Oncogene
PubMed ID:27748757
Resistance to therapeutic antibodies in chronic lymphocytic leukaemia (CLL) is common. In this study, we show that therapeutic antibodies against CD62L (CD62L-Ab) or CD20 (obinutuzumab) were able to induce antibody-dependent cell-mediated cytotoxicity (ADCC) and phagocytosis (ADP) in primary cultures of CLL cells. CLL cells derived from patients with active disease ... More
PI(3,5)P2 biosynthesis regulates oligodendrocyte differentiation by intrinsic and extrinsic mechanisms.
Authors:Mironova YA, Lenk GM, Lin JP, Lee SJ, Twiss JL, Vaccari I, Bolino A, Havton LA, Min SH, Abrams CS, Shrager P, Meisler MH, Giger RJ
Journal:Elife
PubMed ID:27008179
Proper development of the CNS axon-glia unit requires bi-directional communication between axons and oligodendrocytes (OLs). We show that the signaling lipid phosphatidylinositol-3,5-bisphosphate [PI(3,5)P2] is required in neurons and in OLs for normal CNS myelination. In mice, mutations of Fig4, Pikfyve or Vac14, encoding key components of the PI(3,5)P2 biosynthetic complex, ... More
Novel ecto-tagged integrins reveal their trafficking in live cells.
Authors:Huet-Calderwood C, Rivera-Molina F, Iwamoto DV, Kromann EB, Toomre D, Calderwood DA,
Journal:Nat Commun
PubMed ID:28924207
Integrins are abundant heterodimeric cell-surface adhesion receptors essential in multicellular organisms. Integrin function is dynamically modulated by endo-exocytic trafficking, however, major mysteries remain about where, when, and how this occurs in living cells. To address this, here we report the generation of functional recombinant ß1 integrins with traceable tags inserted ... More