ProLong™ Glass Antifade Mountant
ProLong™ Glass Antifade Mountant
ProLong™ Glass Antifade Mountant
Invitrogen™

ProLong™ Glass Antifade Mountant

ProLong Glass Antifade Mountant provides sample and fluorescence preservation but also exhibits the same RI as glass and immersion oil (1.52) allowing high resolution microscopy imaging of cell and tissue samples up to 150 microns in thickness.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
P369822 mL
P369805 x 2 mL
P3698410 mL
Catalog number P36982
Price (USD)
97.65
Online exclusive
107.00
Save 9.35
Each
Quantity:
2 mL
Price (USD)
97.65
Online exclusive
107.00
Save 9.35
Each

ProLong Glass Antifade Mountant is a glycerol-based, hard-setting, ready-to-use mounting media that can be applied directly to fluorescently labeled cells or tissue samples on microscope slides or coverslips. ProLong Glass is compatible with a wide array of fluorescent dyes including Alexa Fluor, FITC, TRTC, Texas Red, Cy3, and Cy5, or proteins (GFP, RFP, mCherry) and with any cell/tissue type for image depths ranging from up to 150 μm for bright, high-resolution Z-Stack, 3D, and 2D images. ProLong Glass mounting media is recommended for all immersion oil imaging applications.

ProLong Glass Antifade Mountant is designed to provide unparalleled photobleach protection across the visible and near infra-red spectra. Prolong Glass Mountant has a refractive index (RI) of 1.52 after curing, similar to glass coverslips, compatible with immersion oil, and oil-immersion microscope optics to enable superior resolution and sensitivity. ProLong Glass Antifade Mountant eliminates the need to use traditional acrylic resins (i.e., fingernail polish), which can damage microscope objectives.

ProLong Glass Antifade Mountant benefits:

  • Up to 75% improvement in axial resolution for sharp images
  • 3–4 times more imageable focal depth compared to mountants with a 1.47 RI, with sharp images for monolayers of cells, tissue slices, or 3D cell cultures up to 150 μm thickness/focal depth
  • Protects fluorescent proteins or dyes from photobleaching

ProLong Glass Antifade Mountant comes ready-to-use in a 2 mL dropper bottle or 10 mL bottle. After removing excess water, just apply a drop of mountant to the sample, add a coverslip, cure, and image. ProLong Glass does not discolor or shrink when cured, making it possible to create high-quality fluorescent images. ProLong Glass is a hard-setting, curing mountant that minimizes optical refraction and spherical aberration in the optical path. The RI determines how much the path of light is bent, or refracted, when entering a material. In microscopy, refractive index differences between specimen and mounting medium should be minimized and can be reduced by using mountants with a high RI such as ProLong Glass mounting media.

Prevent photobleaching across the visible and near-IR spectra
Compatible with almost all fluorescent dyes and fluorescent proteins, ProLong Glass provides superior protection against photobleaching. ProLong Glass mountant does not quench fluorophores and provides high-resolution images with a wide array of fluorescent dyes and proteins. ProLong Glass Antifade Mountant is an ideal reagent for archiving samples and for preserving signal strength during prolonged imaging sessions, such as repeated scanning under confocal laser scanning illumination or with super-resolution microscopy. To image your sample immediately, use our SlowFade Glass Antifade Mountant, a non-curing mountant with similar 1.52 refractive index properties.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
DescriptionProLong™ Glass Antifade Mountant, 1 x 2 mL
Quantity2 mL
Shipping ConditionApproved for shipment on Wet or Dry Ice
Product LineProLong
Product TypeAntifade Mountant
Reagent TypeAntifade Solution
Volume (Metric)2 mL
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C. For long-term storage, store at -5° to -30°C. Avoid freeze/thaw cycles.

Frequently asked questions (FAQs)

Can I use ProLong Glass Antifade Mountant (Cat. No. P36980, P36982, P36984) with thick floating brain sections?

ProLong Glass Antifade Mountant (Cat. No. P36980, P36982, P36984) is compatible with any cell or tissue type, ranging in thickness from 0µm to 150 µm, for producing bright, high-resolution Z-stack, 3-D, and 2-D images.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I do not have epoxy or VALAP to seal the coverslip. Do you offer an alternative coverslip sealant?

We offer ProLong Coverslip Sealant (Cat. No. P56128) that can be used to seal the edges of the coverslip and is compatible with both curing and non-curing mountant. The sealant is easy to apply and is brushed on after the mountant has cured, for long preservation of slides. The product page can be found here.

When using a hard-curing mountant, such as ProLong Antifade Mountant, make sure the mountant is fully cured before applying ProLong Coverslip Sealant. Since this sealant can seal moisture under the coverslip, it can interfere with the curing process.

When using a non-curing mountant, such as SlowFade Antifade Mountants, sealing can take place immediately after mounting.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What do ProLong Gold, ProLong Diamond and ProLong Glass antifade mountant reagents contain?

They are composed of proprietary antifades and polymers in aqueous buffers.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I use ProLong Gold, ProLong Diamond, or ProLong Glass mounting medium in a multi-well plate?

No. If the sample cannot be sealed with a coverslip, we recommend the use of SlowFade antifade mountant.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What can affect sample curing of ProLong Gold-, ProLong Diamond-, and ProLong Glass-mounted samples?

Our ProLong Gold, ProLong Diamond, and ProLong Glass mounting media harden ('cure') by the evaporation of water. Placing the mounted samples in a humid or refrigerated environment would slow down the evaporation process ('curing').

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (60)

Citations & References
Abstract
Light Affects Mood and Learning through Distinct Retina-Brain Pathways.
Authors:Fernandez DC, Fogerson PM, Lazzerini Ospri L, Thomsen MB, Layne RM, Severin D, Zhan J, Singer JH, Kirkwood A, Zhao H, Berson DM, Hattar S
Journal:Cell
PubMed ID:30173913
Light exerts a range of powerful biological effects beyond image vision, including mood and learning regulation. While the source of photic information affecting mood and cognitive functions is well established, viz. intrinsically photosensitive retinal ganglion cells (ipRGCs), the central mediators are unknown. Here we reveal that the direct effects of ... More
Comprehensive structure and functional adaptations of the yeast nuclear pore complex.
Authors:Akey CW, Singh D, Ouch C, Echeverria I, Nudelman I, Varberg JM, Yu Z, Fang F, Shi Y, Wang J, Salzberg D, Song K, Xu C, Gumbart JC, Suslov S, Unruh J, Jaspersen SL, Chait BT, Sali A, Fernandez-Martinez J, Ludtke SJ, Villa E, Rout MP
Journal:Cell
PubMed ID:34982960
Nuclear pore complexes (NPCs) mediate the nucleocytoplasmic transport of macromolecules. Here we provide a structure of the isolated yeast NPC in which the inner ring is resolved by cryo-EM at sub-nanometer resolution to show how flexible connectors tie together different structural and functional layers. These connectors may be targets for ... More
Obesity Shapes Metabolism in the Tumor Microenvironment to Suppress Anti-Tumor Immunity.
Authors:Ringel AE, Drijvers JM, Baker GJ, Catozzi A, García-Cañaveras JC, Gassaway BM, Miller BC, Juneja VR, Nguyen TH, Joshi S, Yao CH, Yoon H, Sage PT, LaFleur MW, Trombley JD, Jacobson CA, Maliga Z, Gygi SP, Sorger PK, Rabinowitz JD, Sharpe AH, Haigis MC
Journal:Cell
PubMed ID:33301708
Obesity is a major cancer risk factor, but how differences in systemic metabolism change the tumor microenvironment (TME) and impact anti-tumor immunity is not understood. Here, we demonstrate that high-fat diet (HFD)-induced obesity impairs CD8(+) T cell function in the murine TME, accelerating tumor growth. We generate a single-cell resolution ... More
Pathogenic ubiquitination of GSDMB inhibits NK cell bactericidal functions.
Authors:Hansen JM, de Jong MF, Wu Q, Zhang LS, Heisler DB, Alto LT, Alto NM
Journal:Cell
PubMed ID:34022140
Gasdermin B (GSDMB) belongs to a large family of pore forming cytolysins that execute inflammatory cell death programs. While genetic studies have linked GSDMB polymorphisms to human disease, its function in the immunological response to pathogens remains poorly understood. Here, we report a dynamic host-pathogen conflict between GSDMB and the ... More
WNK kinases sense molecular crowding and rescue cell volume via phase separation.
Authors:Boyd-Shiwarski CR, Shiwarski DJ, Griffiths SE, Beacham RT, Norrell L, Morrison DE, Wang J, Mann J, Tennant W, Anderson EN, Franks J, Calderon M, Connolly KA, Cheema MU, Weaver CJ, Nkashama LJ, Weckerly CC, Querry KE, Pandey UB, Donnelly CJ, Sun D, Rodan AR, Subramanya AR
Journal:Cell
PubMed ID:36318922
When challenged by hypertonicity, dehydrated cells must recover their volume to survive. This process requires the phosphorylation-dependent regulation of SLC12 cation chloride transporters by WNK kinases, but how these kinases are activated by cell shrinkage remains unknown. Within seconds of cell exposure to hypertonicity, WNK1 concentrates into membraneless condensates, initiating ... More