ProLong™ Gold Antifade Mountant with SYTOX™ Deep Red - FAQs
ProLong™ Gold Antifade Mountant with SYTOX™ Deep Red - FAQs
View additional product information for ProLong™ Gold Antifade Mountant with SYTOX™ Deep Red - FAQs (P36986)
3 product FAQs found
What do ProLong Gold, ProLong Diamond and ProLong Glass antifade mountant reagents contain?
They are composed of proprietary antifades and polymers in aqueous buffers.
Can I use ProLong Gold, ProLong Diamond, or ProLong Glass mounting medium in a multi-well plate?
No. If the sample cannot be sealed with a coverslip, we recommend the use of SlowFade antifade mountant.
What can affect sample curing of ProLong Gold-, ProLong Diamond-, and ProLong Glass-mounted samples?
Our ProLong Gold, ProLong Diamond, and ProLong Glass mounting media harden ('cure') by the evaporation of water. Placing the mounted samples in a humid or refrigerated environment would slow down the evaporation process ('curing').