Electroporation Cuvettes, 0.1 cm
Invitrogen™

Electroporation Cuvettes, 0.1 cm

Electroporation uses short, high-voltage electric pulses to overcome the cell membrane and introduce DNA directly into cells. These cuvettes are자세히 알아보기
Have Questions?
카탈로그 번호수량
P4105050 each
카탈로그 번호 P41050
제품 가격(KRW)
388,000
온라인 행사
Ends: 30-Sep-2026
443,000
할인액 55,000 (12%)
Each
수량:
50 each
제품 가격(KRW)
388,000
온라인 행사
Ends: 30-Sep-2026
443,000
할인액 55,000 (12%)
Each

Electroporation uses short, high-voltage electric pulses to overcome the cell membrane and introduce DNA directly into cells. These cuvettes are specifically designed help achieve high DNA transformation efficiency during the introduction of DNA into bacterial, yeast, fungal, and mammalian cells.

Our electroporation cuvettes are precision manufactured from the highest quality materials to ensure optimal and reproducible transformation results.

Electroporation cuvette features
• Compatible with virtually all common electroporation devices
• Gamma-irradiated and individually packaged to ensure sterility
• Color-coded snap cap for easy identification of gap size (white cap for 0.1 cm cuvette)

Note: Electroporation cuvettes are recommended for use with all of our electroporation competent cells.

For Research Use Only. Not for use in diagnostic procedures.
사양
직경(미터법)0.1 cm
용도(장비)Electroporation Device
제품 유형Electroporation Cuvette
수량50 each
배송 조건Room Temperature
Unit SizeEach
구성 및 보관
• 50 individually wrapped, sterile cuvettes

Store at room temperature.

자주 묻는 질문(FAQ)

Will Invitrogen cuvettes fit my electroporator?

Our cuvettes are known as the "Potter-style" cuvettes, and they fit most electroporators that accept a standard size cuvette.

How can I avoid arcing during electroporation of E. coli?

When electroporating cells at high voltage in conductive buffers, arcing may occur. MgCl2 and PO4 in particular are very conductive.

Some suggestions:

1) Keep ionic strength of cloning reactions to a minimum. If reaction buffers contain high salt, dilute DNA sample in water or TE buffer before electroporation.

2) Minimize addition of conductive ions. The volume of DNA solution should not exceed 5% of the total reaction. Example: 2 ?l DNA per 40 ?l of cells.

3) Be sure no air bubbles are present.

4) Make sure the electrical contacts are clean and tight. Wipe away any condensation on the outside of the cuvette.

5) For best results, the cells should be aliquoted into the bottom of the gap - tap the cuvette gently to help the cells settle to the bottom.

What is meant by "field strength" (kV/cm) when determining settings for electroporation? What are typical settings for different cell types?

The two most important electrical parameters for consideration in electroporation are pulse length and field strength. Field strength is defined as volts/centimeter, where V is equal to the initial peak voltage and cm is equal to the measurement of the gap between the electrodes of the cuvette in centimeters. For example, during electroporation of bacteria at 1,500 Volts in a 0.1 cm cuvette, the field strength would be 15,000 volts/cm, or 15 kV/cm. Typically, electroporation of bacteria requires field strengths of greater than 15 kV/cm. Yeast cells require 6-8 kV/cm, and mammalian cells often require optimization between 0.5 to 2.5 kV/cm.

What is the maximum volume of E. coli (bacterial) cells that can be electroporated in a 0.1 cm cuvette?

20-80 ul is a general suggested volume, but follow the optimized protocols for the cells and cuvettes that you are using.

인용 및 참조 문헌 (8)

인용 및 참조 문헌
Abstract
Transfection techniques for producing recombinant baculoviruses.
Authors:Trotter KM,Wood HA
Journal:Molecular biotechnology
PubMed ID:9067978
Synthesis and anticonvulsant and neurotoxic properties of substituted N-phenyl derivatives of the phthalimide pharmacophore.
Authors:Vamecq J, Bac P, Herrenknecht C, Maurois P, Delcourt P, Stables JP
Journal:J Med Chem
PubMed ID:10753468
'A series of compounds including 4-amino (1), 3-amino (2), 4-nitro (3), 2-methyl-3-amino (4), 2-methyl-3-nitro (5), 2-methyl-4-amino (6), 2-methyl-4-nitro (7), 2-methyl-5-amino (8), 2-methyl-5-nitro (9), 2-methyl-6-amino (10), 2-methyl-6-nitro (11), 2,6-dimethyl (12), 2-methyl-3-carboxy (13), 2-methoxycarbonyl (14), 2-methyl-4-methoxy (15), 2,4-dimethoxy (16), 2-chloro-4-amino (17), and 2-chloro-4-nitro (18) N-phenyl substituents of phthalimide were evaluated along with ... More
Optimization of the transfection of human endothelial cells by electroporation.
Authors:Schwachtgen JL, Ferreira V, Meyer D, Kerbiriou-Nabias D
Journal:Biotechniques
PubMed ID:7840968
A method for efficient transfection of human umbilical vein endothelial cells (HUVEC) is presented here. The procedure involves cell synchronization followed by electroporation and allows the detection of the activity of low-strength promoters. Sodium butyrate, added to the culture medium after electroporation, strongly potentiates the transcriptional activity of viral promoters. ... More
Expression of mRNA electroporated into plant and animal cells.
Authors: Callis J; Fromm M; Walbot V;
Journal:Nucleic Acids Res
PubMed ID:3475678
A general method to introduce RNA molecules into plant protoplasts and animal cells is described. This technique utilizes the ability of electric pulses of high field strength to form pores in biomembranes. RNA molecules containing the coding region for the bacterial enzyme chloramphenicol acetyltransferase (CAT) were used as a model ... More
Initiation of the phosphocholine-specific response to Proteus morganii. B cell transfectants expressing unmutated VH/VL can respond to stimulation by P. morganii antigen.
Authors:Penner SJ, George J, Claflin L
Journal:J Immunol
PubMed ID:7544373
We wished to resolve a paradox of how the response to the phosphocholine (PC) determinant of Proteus morganii could be initiated from a precursor B cell whose receptor, in unmutated form as Ab, appears to be unable to bind Ag. Unmutated VH and unmutated VL constructs were co-transfected into the ... More