Electroporation Cuvettes, 0.2 cm
Invitrogen™

Electroporation Cuvettes, 0.2 cm

Electroporation uses short, high-voltage electric pulses to overcome the cell membrane and introduce DNA directly into cells. These cuvettes are자세히 알아보기
Have Questions?
카탈로그 번호수량
P4505050 each
카탈로그 번호 P45050
제품 가격(KRW)
377,000
온라인 행사
Ends: 30-Jun-2026
430,000
할인액 53,000 (12%)
Each
수량:
50 each
제품 가격(KRW)
377,000
온라인 행사
Ends: 30-Jun-2026
430,000
할인액 53,000 (12%)
Each

Electroporation uses short, high-voltage electric pulses to overcome the cell membrane and introduce DNA directly into cells. These cuvettes are specifically designed help achieve high DNA transformation efficiency during the introduction of DNA into bacterial, yeast, fungal, and mammalian cells.

Our electroporation cuvettes are precision manufactured from the highest quality materials to ensure optimal and reproducible transformation results.

Electroporation cuvette features
• Compatible with virtually all common electroporation devices
• Gamma-irradiated and individually packaged to ensure sterility
• Color-coded snap cap for easy identification of gap size (blue cap for 0.2 cm cuvette)

Note: Electroporation cuvettes are recommended for use with all of our electroporation competent cells.

For Research Use Only. Not for use in diagnostic procedures.
사양
직경(미터법)0.2 cm
용도(장비)Electroporation Device
제품 유형Electroporation Cuvette
수량50 each
배송 조건Room Temperature
Unit SizeEach
구성 및 보관
• 50 individually wrapped, sterile cuvettes

Store at room temperature.

자주 묻는 질문(FAQ)

Will Invitrogen cuvettes fit my electroporator?

Our cuvettes are known as the "Potter-style" cuvettes, and they fit most electroporators that accept a standard size cuvette.

How can I avoid arcing during electroporation of E. coli?

When electroporating cells at high voltage in conductive buffers, arcing may occur. MgCl2 and PO4 in particular are very conductive.

Some suggestions:

1) Keep ionic strength of cloning reactions to a minimum. If reaction buffers contain high salt, dilute DNA sample in water or TE buffer before electroporation.

2) Minimize addition of conductive ions. The volume of DNA solution should not exceed 5% of the total reaction. Example: 2 ?l DNA per 40 ?l of cells.

3) Be sure no air bubbles are present.

4) Make sure the electrical contacts are clean and tight. Wipe away any condensation on the outside of the cuvette.

5) For best results, the cells should be aliquoted into the bottom of the gap - tap the cuvette gently to help the cells settle to the bottom.

What is meant by "field strength" (kV/cm) when determining settings for electroporation? What are typical settings for different cell types?

The two most important electrical parameters for consideration in electroporation are pulse length and field strength. Field strength is defined as volts/centimeter, where V is equal to the initial peak voltage and cm is equal to the measurement of the gap between the electrodes of the cuvette in centimeters. For example, during electroporation of bacteria at 1,500 Volts in a 0.1 cm cuvette, the field strength would be 15,000 volts/cm, or 15 kV/cm. Typically, electroporation of bacteria requires field strengths of greater than 15 kV/cm. Yeast cells require 6-8 kV/cm, and mammalian cells often require optimization between 0.5 to 2.5 kV/cm.

What are recommended settings for bacterial electroporation of 80 ul cells in 0.2 cm cuvettes? What is the maximum cell volume to use in a 0.2 cm cuvette?

Depending upon the manufacturer of the electroporator, the settings may range in value (25 -50 ?F, 100-200 ohms, 1500-2500 volts). Follow the manufacturer's recommended settings for bacterial electroporation. The maximum recommended cell volume that should be used for a 0.2 cm cuvette without compromising efficiency is 160 ?l.

인용 및 참조 문헌 (5)

인용 및 참조 문헌
Abstract
Expression cloning and functional characterization of human cDNA for ganglioside GM3 synthase.
Authors:Ishii A, Ohta M, Watanabe Y, Matsuda K, Ishiyama K, Sakoe K, Nakamura M, Inokuchi J, Sanai Y, Saito M
Journal:J Biol Chem
PubMed ID:9822625
'Ganglioside GM3 is a major glycosphingolipid in the plasma membrane and is widely distributed in vertebrates. We describe here the isolation of a human cDNA whose protein product is responsible for the synthesis of GM3. The cloned cDNA spanned 2,359 base pairs, with an open reading frame encoding a protein ... More
Introduction of deoxyribonucleoside triphosphates into intact cells by electroporation.
Authors: Sokoloski J A; Jastreboff M M; Bertino J R; Sartorelli A C; Narayanan R;
Journal:Anal Biochem
PubMed ID:3812972
A simple method, employing high-voltage electric discharge (electroporation), was developed to introduce phosphorylated nucleosides into the cytoplasm of viable cells. HL-60 leukemia cells permeabilized by this technique remained viable and incorporated deoxyribonucleoside triphosphates into nuclear DNA. Furthermore, DNA synthesis was depressed for at least 24 h in HL-60 cells made ... More
T cell receptor (TCR) mini-gene mRNA expression regulated by nonsense codons: a nuclear-associated translation-like mechanism
Authors:Li S, Leonard D, Wilkinson MF
Journal:J Exp Med
PubMed ID:9091590
Premature termination codons (PTCs) are known to decrease mRNA levels. Here, we report our investigation of the mechanism for this downregulation using the TCR-beta gene, which acquires PTCs as a result of programmed rearrangements that occur during normal thymic development. We found that a mini-gene version of this gene, which ... More
Expression of a human mutant monocyte chemotactic protein 3 in Pichia pastoris and characterization as an MCP-3 receptor antagonist.
Authors:Masure S, Paemen L, Proost P, Van Damme J, Opdenakker G
Journal:J Interferon Cytokine Res
PubMed ID:8590307
The cDNA encoding human monocyte chemotactic protein 3 (hMCP-3) was cloned in pHIL-S1, a vector designed for inducible secreted heterologous expression in the methylotrophic yeast Pichia pastoris. After transformation of P. pastoris by electroporation, several clones with the human MCP-3 gene integrated at the alcohol oxidase (AOX-1) locus were isolated. ... More
Short technical reports. Effects of lipopolysaccharide on transfection efficiency in eukaryotic cells.
Authors: Weber M; Möller K; Welzeck M; Schorr J;
Journal:Biotechniques
PubMed ID:8747659
The suitability of different purification methods for preparation of plasmid DNA for transfection into eukaryotic cells was systematically investigated. The reporter plasmid, pRSVcat, was prepared using several methods, and residual impurities in the preparations were quantitated. Transfection with these preparations was performed with several cell lines (HeLa, Huh7, COS7 and ... More