PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents
Invitrogen™

PrestoBlue™ and PrestoBlue™ HS Cell Viability Reagents

Monitor cell proliferation and viability with the resazurin-based PrestoBlue and PrestoBlue HS cell viability reagents. These nontoxic and cell permeable reagents quantitatively measure cell viability in as little as 10 minutes using absorbance- or fluorescence-based microplate readers.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPurity or Quality Grade
P50201100 mLHS Purified
A1326125 mLStandard
A13262100 mLStandard
P5020025 mLHS Purified
Catalog number P50201
Price (USD)
744.65
Online exclusive
768.00
Save 23.35
Each
Quantity:
100 mL
Purity or Quality Grade:
HS Purified
Price (USD)
744.65
Online exclusive
768.00
Save 23.35
Each
PrestoBlue and PrestoBlue HS cell viability reagents are ready-to-use, nontoxic, resazurin-based solutions that function as long term kinetic cell health indicators for fluorescence and absorbance-based microplate assays. These reagents are cell permeable and use the reducing power of living cells to quantitatively measure cell proliferation and viability in as little as 10 minutes. PrestoBlue HS Cell Viability Reagent is an improved version of PrestoBlue reagent, containing highly purified resazurin, which increases sensitivity and reduces background in cell proliferation assays. Add the power of multiplexing with reagents such as the CyQUANT Direct assay to improve your data analysis.

Measuring changes in cell viability is a fundamental method for assessing cell health, determining genotoxicity, and evaluating anti-cancer drugs. Cell health can be monitored by detecting changes in several key indicators, including changes to plasma membrane integrity, DNA synthesis, DNA content, enzyme activity, presence of ATP, and cellular reducing environment.

Features of PrestoBlue and PrestoBlue HS cell viability reagents include:

  • Optimized for reliable detection of mammalian cell viability in only 10 minutes
  • Measure cell viability and proliferation in various cell types, including mammalian (2D and 3D), bacteria, and yeast cells
  • A convenient add-and-read format—no mixing, washing, or cell lysis required
  • Compatible with either fluorescence- or absorbance-based microplate readers
  • Obtain high quality cell proliferation results with a large dynamic range
  • Monitor living cells using real time kinetics and continue culturing them for downstream multiplexed functional assays

Quick 10-minute incubations

Detect mammalian cell viability after just a 10-minute incubation with PrestoBlue and PrestoBlue HS cell viability reagents. These are the only live-cell viability reagents on the market that provide robust data in 10 minutes following reagent addition. All other resazurin-based cell viability reagents require a 1–4 hour incubation with the reagent prior to obtaining data. For best results using 3D samples, it is recommended to incubate the samples for 18–24 hours prior to measurements.

Exceptional sensitivity and accuracy

PrestoBlue and PrestoBlue HS cell viability reagents can detect as few as 12 cells per well in a 384-well plate, depending on the cell type and incubation time. After a 10-minute incubation, PrestoBlue reagent can linearly detect cells down to 98 cells/well with a Z' value greater than 0.5.

Simplified assay format

PrestoBlue and PrestoBlue HS cell viability reagents are addition-only reagents, eliminating the need for cell lysis or solubilization steps required by other assays. This minimizes hands-on time and simplifies the process.

Nontoxic live cell assays

PrestoBlue and PrestoBlue HS cell viability reagents are nontoxic and do not require cell lysis, allowing cells to be incubated for up to 24 hours. Since no lysis is required, the diluted PrestoBlue or PrestoBlue HS solution can be removed from the cells and replaced with complete growth medium for further culturing and use in downstream assays.

PrestoBlue HS reagent—a highly purified version of PrestoBlue reagent with improved performance

All resazurin-based reagents contain a detectable amount of resorufin contamination due to the synthesis and manufacturing process, which contributes to high background fluorescence and reduces performance in cell viability assays. To reduce background and improve performance, an innovative purification process was developed to remove the contaminating resorufin, resulting in the highly pure resazurin used in the PrestoBlue HS Cell Viability Reagent. This highly purified, nontoxic resazurin reduces background fluorescence by >50% and increases signal-to-background by >100% in cell viability microplate assays.

Features of PrestoBlue HS Cell Viability Reagent include:

  • Removal of contaminants from resazurin, resulting in >50% reduction in background fluorescence
  • Increased signal-to-background ratio by >100%, providing a large assay signal window
  • Offers high sensitivity with a linear response, detecting as few as 10 cells per well

Mechanism of PrestoBlue and PrestoBlue HS reagents

Using resazurin-based reagents to monitor changes in cellular reducing environment or metabolic activity is a well-established and reliable method to indicate cell viability or death. PrestoBlue and PrestoBlue HS reagents contain resazurin, which is a nontoxic, cell permeable compound that is blue in color and virtually nonfluorescent. When added to cells, the resazurin-based PrestoBlue and PrestoBlue HS reagents enter cells and are modified by the reducing environment of viable cells. In live cells, resazurin is reduced to resorufin, a compound that is red in color and highly fluorescent. Viable cells continuously convert resazurin to resorufin, increasing the overall fluorescence of the medium surrounding the cells. The conversion of resazurin to resorufin also results in a pronounced color change, allowing cell viability to be detected using either absorbance- or fluorescence-based plate readers.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell PermeabilityCell-permeant
Cell TypeMammalian Cells
Detection MethodAbsorbance, Fluorescence
Dye TypeResorufin
Emission Wavelength590
Excitation Wavelength560 nm
FormLiquid
Format96-well Plate, 384-well Plate, Cuvettes
Purity or Quality GradeHS Purified
Quantity100 mL
Shipping ConditionWet Ice
Absorbance Wavelength570 nm
ColorRed
For Use With (Application)Cell Viability
For Use With (Equipment)Microplate Reader, Spectrophotometer, HTS Reader
Product LinePrestoBlue
Product TypeCell Viability Reagent
Reference Wavelength600 nm
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C and protect from light.

Frequently asked questions (FAQs)

Can I combine Invitrogen CellEvent Caspase-3/7 Green Detection Reagent with PrestoBlue Cell Viability Reagent?

Yes. Invitrogen CellEvent Caspase-3/7 Green Detection Reagent has been demonstrated in combination with Invitrogen PrestoBlue Cell Viability Reagent in the same sample. CellEvent Caspase reagent fluorescence reports activated caspase, while PrestoBlue Cell Viability Reagent reports cellular metabolic activity as an indicator of viability. Follow the instructions for each reagent and use single-reagent controls to evaluate signal interference.

What viability or cytotoxicity reagents would you recommend for a high throughput assay with multiple time points?

We recommend alamarBlue HS Cell Viability Reagent (Cat. No. A50101) or prestoBlue HS Cell Viability Reagent (Cat. Nos. P50200, P50201) for experiments with multiple time points or longer incubation time. We recommend that you add the reagent at each time-point at the end of each incubation period rather than continuous measurement. This prevents the reagent from being converted by metabolically active cells, which could confound results.

Use the link below to help you decide which reagent is most suitable for your purpose:

Microplate Assays for Cell Viability

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Why would I use PrestoBlue HS Cell Viability Reagent instead of the original PrestoBlue Cell Viability Reagent?

PrestoBlue HS Cell Viability Reagent provides a lower background, suitable for analyzing a lower number of cells per well. Also, the removal of impurities in the 'HS' reagent limits any artifacts that may be critical to your cells' viability.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Does PrestoBlue HS Cell Viability Reagent have the same properties as the original PrestoBlue Cell Viability Reagent?

Yes and in addition, PrestoBlue HS Cell Viability Reagent has a higher purity which results in a lower background.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What is the difference between PrestoBlue HS Cell Viability Reagent (Cat. Nos. P50200, P50201) and the original PrestoBlue Cell Viability Reagent (Cat. Nos. A13261, A13262)?

Both products utilize resazurin as the indicator dye. PrestoBlue HS Cell Viability Reagent undergoes a purification process that removes any residual resorufin that may be present from the standard manufacturing process for resazurin. This purification results in a product that has very low background. PrestoBlue HS Cell Viability Reagent may be used on samples with as few as 10 cells/well, compared to a minimum of 12 cells/well for the orginal PrestoBlue Cell Viability Reagent.
Note: Minimum cell number/well is also dependent upon reducing activity of the cell type. Results may vary.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.