E-Shot™ Standard Electroporation Cuvettes, 0.2 cm - FAQs
E-Shot™ Standard Electroporation Cuvettes, 0.2 cm - FAQs
View additional product information for E-Shot™ Standard Electroporation Cuvettes, 0.2 cm - FAQs (P52050)
1 product FAQs found
Will Invitrogen cuvettes fit my electroporator?
Our cuvettes are known as the "Potter-style" cuvettes, and they fit most electroporators that accept a standard size cuvette.