Get upto 200X more sensitivity in ICC/IHC/FISH for more clarity and definition with new Tyramide SuperBoost™ kits
Protein-Protein Crosslinking Kit
Catalog number:  P6305
Invitrogen™  Related applications: Labeling Chemistry | Protein Labeling & Crosslinking
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Protein-Protein Crosslinking Kit provides all of the reagents and purification media required to perform three protein-protein conjugations in which neither protein contains available thiol residues. Briefly, the amines of one of the proteins is thiolated with SPDP and the amines of the second protein are converted to thiol-reactive maleimides with SMCC. This kit was designed to prepare and purify protein-protein conjugates, including crosslinking phycobiliproteins (R-PE) to antibodies, but it can be modified to generate peptide-protein or enzyme-nucleic acid conjugates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Labeling Scale: 0.2-3 mg
Labeling Target: Proteins (General)
Product Size: 1 kit
Shipping Condition: Room Temperature
Contents & storage
Store in refrigerator (2–8°C).
Documents
Manuals & protocols
Recommended products