Get the sharpest, brightest images. Try new Invitrogen™ SlowFade™ Glass antifade RIMS mountant with refractive index of 1.52. Learn more »
Citations & References (56)
Invitrogen™
Phospholipase C Protein, Phosphatidylinositol-Specific (from Bacillus cereus)
This phosphatidylinositol-specific phospholipase C is a highly purified enzyme preparation from Bacillus cereus which cleaves phospholipid phosphatidylinositol (PI) into 2Read more
Have Questions?
Catalog Number
Quantity
P6466
50 μL
Catalog number P6466
Price (USD)
210.00
Each
Quantity:
50 μL
Price (USD)
210.00
Each
This phosphatidylinositol-specific phospholipase C is a highly purified enzyme preparation from Bacillus cereus which cleaves phospholipid phosphatidylinositol (PI) into 2 molecules. This preparation is suitable for both enzymology and cell biology applications.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration100 U⁄ml
Protein SubtypeOther Proteins
Quantity50 μL
Research CategorySignal Transduction
Shipping ConditionWet Ice
SourceBacillus cereus
For Use With (Application)Assay Standard
FormSolution
SpeciesB. cereus
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).
Citations & References (56)
Citations & References
Abstract
Mechanism and structure based inhibitors of phospholipase C enzymes.
Authors:Roberts MF, Wu Y, Zhou C, Geng D, Tan C
Journal:Adv Enzyme Regul
PubMed ID:8869740
'PI-specific PLC enzymes are a key component of phosphatidylinositol-mediated signaling pathways since the hydrophobic product, diacylglycerol, activates protein kinase C and the water-soluble product, inositol trisphosphate, is involved in Ca2+ mobilization. Nonspecific, or PC-PLC, enzymes can generate diacylglycerol without Ca2+ mobilization. A series of inhibitors, both lipophilic and water-soluble, have ... More
Diacylglycerol induces fusion of nuclear envelope membrane precursor vesicles.
'Purified membrane vesicles isolated from sea urchin eggs form nuclear envelopes around sperm nuclei following GTP hydrolysis in the presence of cytosol. A low density subfraction of these vesicles (MV1), highly enriched in phosphatidylinositol (PtdIns), is required for nuclear envelope formation. Membrane fusion of MV1 with a second fraction that ... More
A turbidometric assay for phospholipase C and sphingomyelinase.
Authors:Torley L, Silverstrim C, Pickett W
Journal:Anal Biochem
PubMed ID:7864373
'We describe a simple turbidometric assay for phosphatidylcholine-specific phospholipase C (PC-PLC) (EC 3.1.4.3), phosphatidylinositol-specific phospholipase C (PI-PLC) (EC 3.1.4.10), and sphingomyelinase (SMase) (EC 3.1.4.12), suitable for high-volume screening using unmodified substrates. Under the conditions described, 1 to 10 U/ml of PC-PLC (Bacillus cereus) induces a rapid and continuous increase in ... More
Design, synthesis, and evaluation of water-soluble phospholipid analogues as inhibitors of phospholipase C from Bacillus cereus.
Authors:Franklin CL, Li H, Martin SF
Journal:J Org Chem
PubMed ID:12968879
'The rate of hydrolysis of natural phospholipids by the phosphatidylcholine-preferring phospholipase C from Bacillus cereus (PLC(Bc)) follows the order phosphatidylcholine > phosphatidylethanolamine >> phosphatidyl-l-serine. To probe the structural basis for this substrate specificity, a series of water-soluble, nonhydrolyzable substrate analogues were needed so their complexes with the enzyme could be ... More
Internalization of exogenously added memapsin 2 (beta-secretase) ectodomain by cells is mediated by amyloid precursor protein.
'Memapsin 2 (beta-secretase) is the protease that initiates cleavage of amyloid precursor protein (APP) leading to the production of amyloid-beta (Abeta) peptide and the onset of Alzheimer''s disease. Both APP and memapsin 2 are Type I transmembrane proteins and are endocytosed into endosomes where APP is cleaved by memapsin 2. ... More