What is a suitable alternative for the Protein Molecular Weight Standards (broad range) (Cat. No. P6649)?
A suitable alternative for the Protein Molecular Weight Standards (broad range) (Cat. No. P6649) is the PageRuler Unstained Broad Range Protein Ladder (Cat. No. 26630). The PageRuler Unstained Broad Range Protein Ladder is a mixture of 11 proteins (5 to 250 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. If used for fluorescence application, we recommend using a lower amount. i.e., start with 1 µL. The fluorescence is provided by the gel stain.
Find additional tips, troubleshooting help, and resources within our Protein Standards and Ladders Support Center.
Find additional tips, troubleshooting help, and resources within our Protein Standards and Ladders Support Center.